Glycine, 0.2M buffer soln., pH 3.0, low endotoxin
Glycine, 0.2M buffer soln., pH 3.0, low endotoxin
Glycine, 0.2M buffer soln., pH 3.0, low endotoxin
Glycine, 0.2M buffer soln., pH 3.0, low endotoxin
Thermo Scientific Chemicals

Glycine, 0.2M buffer soln., pH 3.0, low endotoxin

CAS: 56-40-675.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J67349.AK
also known as J67349-AK
Price (EUR)
102,00
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
102,00
Each
Add to cart
Specifications
CAS56-40-6
ColorColorless
Recommended StorageAmbient Temperatures
Concentration0.2M buffer soln.
Molecular Weight (g/mol)75.067
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only