Thermo Scientific Chemicals

Metronidazole, 50 mg/ml in 0.1N HCl, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67362.AD
50 mL, Each
Thermo Scientific Chemicals

Metronidazole, 50 mg/ml in 0.1N HCl, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67362.AD
50 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67362.AD
also known as J67362-AD
Price (USD)
Price: 188.00
Online price: 162.65
Your price:
Quantity
-

Chemical Identifiers

CAS
443-48-1
IUPAC Name
2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethan-1-ol
Molecular Formula
C6H9N3O3
InChI Key
VAOCPAMSLUNLGC-UHFFFAOYSA-N
SMILES
CC1=NC=C(N1CCO)[N+]([O-])=O
Form
Liquid
Appearance (Color)
Clear
Concentration
50 mg/mL

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text