Thermo Scientific Chemicals

Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67369.XF
1 mL, Each
Thermo Scientific Chemicals

Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67369.XF
1 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67369.XF
also known as J67369-XF
Price (USD)
Price: 57.00
Online price: 48.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1400-61-9
IUPAC Name
(1S,15S,16R,17R,18S,19E,21E,25E,27E,29E,31E)-33-{[(2S,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,4,7,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,25,27,29,31-hexaene-36-carboxylic acid
Molecular Formula
C47H75NO17
InChI Key
VQOXZBDYSJBXMA-RKEBNKJGSA-N
SMILES
C[C@H]1O[C@H](OC2CC3O[C@@](O)(CC(O)C3C(O)=O)CC(O)C(O)CCC(O)CC(O)CC(O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)C=CC=CCCC=CC=CC=CC=C2)[C@@H](O)[C@@H](N)[C@@H]1O
Appearance (Color)
Clear pale yellow to yellow
Form
Liquid, prepared in methanol
Composition
Nystatin: 10 mg/mL

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text