HEPES-buffered saline (2X, pH 7.0), low endotoxin
HEPES-buffered saline (2X, pH 7.0), low endotoxin
Thermo Scientific Chemicals

HEPES-buffered saline (2X, pH 7.0), low endotoxin

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J67505.AK
also known as J67505-AK
Price (EUR)
103,00
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
103,00
Each
Add to cart
Specifications
Chemical Name or MaterialHEPES-buffered saline (2X, pH 7.0)
ColorColorless
Name Notelow endotoxin
Recommended StorageKeep cold
Storage Note 12° to 8°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only