Thermo Scientific Chemicals

Hanks' Balanced Salt Solution (1X), without calcium, without magnesium, without phenol red

Catalog number: J67681.K2
1L, Each
Thermo Scientific Chemicals

Hanks' Balanced Salt Solution (1X), without calcium, without magnesium, without phenol red

Catalog number: J67681.K2
1L, Each
Quantity
Catalog number: J67681.K2
also known as J67681-K2
Price (USD)
Price: 140.00
Online price: 117.65
Your price:
Quantity
-
Comment
Components are dissolved in UV treated 18.2 megohm water, pH adjusted and then filtered through 0.22 micron filter
pH
7.4 ± 0.15
Form
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text