Krebs-Ringer Solution, without calcium
Thermo Scientific Chemicals
Krebs-Ringer Solution, without calcium
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J67839.K2
also known as J67839-K2
Price (EUR)
139,65
Online Exclusive
155,00
Save 15,35 (10%)
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
139,65
Online Exclusive
155,00
Save 15,35 (10%)
Each
Specifications
PackagingPlastic Bottle
Recommended StorageKeep cold
Physical FormLiquid
Quantity1 L
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the pH of the Krebs-Ringer Solution, without calcium?
The pH of the Krebs-Ringer Solution, without calcium is 7.3.
What are the concentrations of the components of Krebs-Ringer Solution, without calcium?
The Krebs-Ringer Solution, without calcium contains 120 mM sodium chloride, 5 mM potassium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5.5 mM D-glucose.