Thermo Scientific Chemicals

Water, RNase-Free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

500 mL, Nalgene bottle, Each
Thermo Scientific Chemicals

Water, RNase-Free, DEPC Treated, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

500 mL, Nalgene bottle, Each
Quantity
Packaging
Catalog number: J70783.AP
Price (USD)
Price: 72.90
Your Price:
Quantity
-

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
DNase
(endo): Passes test
RNase
Passes test
Comment
Contaminant Testing: Enzyme Inhibition: Passes test
DNase
(exo): Passes test

Description

RUO – Research Use Only.