How should I store Proteinase K Lyophilized Powder (Cat. No. J76230.MC)?
Proteinase K Lyophilized Powder should be shipped on cold pack and stored dry at 4 degrees C upon receipt.
Search
View additional product information for Proteinase K Lyophilized Powder - FAQs (J76230.MC)
3 product FAQs found