PCR Markers should be shipped on ice and stored at 4 degrees C for short-term use, or -20 degrees C for long-term storage (for example until end of shelf life).
Note: 6X DNA Loading Buffer (OXG) is included with this product and should be shipped at ambient temperature.