Thermo Scientific™
Pasteurella multocida Toxin (PMT) Detection Kit
Related applications:
Industrial Microbiology
Detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs1, with Thermo Scientific™ Oxoid™ Pasteurella multocida Toxin PMT Kit. The enzyme-linked immunosorbent assay is designed to deliver specific, sensitive, and fast detection of Pasteurella multocida toxin in porcine samples.
Back to top
Progressive atrophic rhinitis is a disease of pigs, caused by infection with toxigenic P. multocida.1 The pathogenesis of the disease is linked to a single, well-characterized component: the Pasteurella multocida toxin (PMT). The clinical manifestations of the disease are shortening or distortion of the snout, sneezing, nasal discharge, and epitaxis. Reduced growth rate is seen in severe cases.
Use ELISA Systems Pasteurella multocida Toxin PMT Detection Kit for the detection of Pasteurella multocida toxin in porcine samples.
  • Simple: Ready-to-use liquid reagents and concentrates make the assay easy to implement and run
  • Sensitive: Detect proteins at low levels for improved assurance of safety
  • Specific: Detect PMT in any type of sample, including extracts of primary mixed cultures obtained from, e.g., nasal swabs or washings2
Other names for the PMT include dermonecrotic toxin (DNT), heat labile toxin (HLT), turbinate atrophy toxin (TAT), and osteolytic toxin.
For Veterinary Use Only.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
1. Pedersen KB, Nielsen JP, Foged NT, Elling F, Nielsen NC, Willeberg P. Atrophic rhinitis in pigs: proposal for a revised definition. Vet Rec 1988; 122: 190-1.
2. Foged NT, Nielsen JP, Pedersen KB. Differentiation of toxigenic from non-toxigenic Pasteurella multocida by enzymelinked immunosorbent assay, Journal of Clinical Microbiology, 1988; 26: 1419-20.