GeneJET Plant Genomic DNA Purification Kit
Thermo Scientific™
GeneJET Plant Genomic DNA Purification Kit
Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomicRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K079150 Preps
K0792250 Preps
Catalog number K0791
Price (USD)
167.65
Online exclusive
233.00
Save 65.35
Each
Quantity:
50 Preps
Price (USD)
167.65
Online exclusive
233.00
Save 65.35
Each
Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.
Highlights
Fast—procedure completed in 30 min following cell lysis
Convenient—protocol does not include clogging-prone lysate filtration step
High yield—up to 32 μg of high quality DNA from 100 mg sample, including nuclear, mitochondrial, and chloroplast DNA
Compatible—use with awide variety of samples (leaves, roots, sprouts and other parts of various plants)
The standard procedure takes less than 30 minutes following cell lysis and yields purified DNA greater than 30 kb in size. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and the age of the tissue sample. The typical yield from a standard tissue source (e. g. young wheat leaves) is 30 to 32 μg from 100 mg of tissue. Isolated DNA can be used directly in PCR, qPCR, southern blotting, and enzymatic reactions.
Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
FastDigest and conventional restriction digestion
• Southern blotting
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeSpin Column
For Use With (Application)Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineGeneJET
Product TypeGenomic DNA Purification Kit
Quantity50 Preps
Sample TypePlants
TargetGenomic DNA
Test Time<20 min.
FormatSpin Column
Isolation TechnologySpin Column
Unit SizeEach
Frequently asked questions (FAQs)
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Can I use a vacuum manifold with GeneJET kits?
The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.
At what temperature should I perform centrifugation steps in the GeneJET kits?
We recommend performing centrifugation steps at room temperature when the temperature is not specified.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.