GeneJET Plant Genomic DNA Purification Kit
Thermo Scientific™
GeneJET Plant Genomic DNA Purification Kit
Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomicRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K0792250 Preps
K079150 Preps
Catalog number K0792
Price (USD)
721.65
Online exclusive
1,002.00
Save 280.35
Each
Quantity:
250 Preps
Price (USD)
721.65
Online exclusive
1,002.00
Save 280.35
Each
Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.
Highlights
• Fast—procedure completed in 30 min following cell lysis
• Convenient—protocol does not include clogging-prone lysate filtration step
• High yield—up to 32 μg of high quality DNA from 100 mg sample, including nuclear, mitochondrial, and chloroplast DNA
• Compatible—use with awide variety of samples (leaves, roots, sprouts and other parts of various plants)
The standard procedure takes less than 30 minutes following cell lysis and yields purified DNA greater than 30 kb in size. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and the age of the tissue sample. The typical yield from a standard tissue source (e. g. young wheat leaves) is 30 to 32 μg from 100 mg of tissue. Isolated DNA can be used directly in PCR, qPCR, southern blotting, and enzymatic reactions.
Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeSpin Column
Elution Volume200 μL
Final Product TypegDNA
For Use With (Application)PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Quantity250 Preps
Sample TypePlants
ScaleMini
Shipping ConditionRoom Temperature
Starting Material Amount≤100 mg of fresh/frozen
≤20 mg lyophilized
Test Time30 min. (After sample homogenization)
Yield≤32 μg per 100 mg tissue
Isolation TechnologySilica Spin Column
Unit SizeEach
Contents & Storage
• 6 x 1 mL RNase A; store at room temperature until opened, –20°C after opening
• 120 mL Lysis Buffer A; room temperature
• 15 mL Lysis Buffer B; room temperature
• 40 mL Precipitation Solution; room temperature
• 120 mL Plant gDNA Binding Solution; room temperature
• 40 mL Wash Buffer I (concentrated); room temperature
• 40 mL Wash Buffer II (concentrated); room temperature
• 150 mL Elution Buffer; room temperature
• 250 GeneJET Purification Columns with Collection Tubes; room temperature
• 250 Collection Tubes; room temperature
Frequently asked questions (FAQs)
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Can I use a vacuum manifold with GeneJET kits?
The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.
At what temperature should I perform centrifugation steps in the GeneJET kits?
We recommend performing centrifugation steps at room temperature when the temperature is not specified.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.