GeneJET NGS Cleanup Kit, 50 Preps - FAQs
GeneJET NGS Cleanup Kit, 50 Preps - FAQs
View additional product information for GeneJET NGS Cleanup Kit - FAQs (K0851)
2 product FAQs found
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.