Will quality of DNA purified with GeneJET FFPE DNA Purification Kit affect downstream applications?
Due to high de-crosslinking efficiency of GeneJET FFPE DNA Purification Kit, high-quality purified DNA can be directly used in sensitive downstream applications such as qPCR, PCR (amplicons up to 1 kb) or stored at -20 degrees C.
What are expected yields from the GeneJET FFPE DNA Purification Kit?
DNA yield and quality from FFPE tissue may vary considerably depending on the tissue source, the thickness of the slice, how much tissue is in the slice, the age of the sample, post-sampling delay before fixation, fixation time, etc. We recover up to 8 µg of genomic DNA from one slice (10 µm thickness) of FFPE sample depending on the tissue type.