Product Image
Invitrogen™
Champion™ pET101 Directional TOPO™ Expression Kit with BL21 Star™ (DE3) One Shot™ Chemically Competent E. coli
The Champion™ pET Expression System yields the highest-level protein production in E. coli. During expression, your protein of interest canRead more
Have Questions?
Catalog NumberQuantity
K1010120 reactions
Catalog number K10101
Price (USD)
1,280.60
Special offer
Online exclusive
Ends: 27-Jun-2026
1,348.00
Save 67.40 (5%)
Each
Quantity:
20 reactions
Price (USD)
1,280.60
Special offer
Online exclusive
Ends: 27-Jun-2026
1,348.00
Save 67.40 (5%)
Each
The Champion™ pET Expression System yields the highest-level protein production in E. coli. During expression, your protein of interest can reach levels greater than 50 percent of total cellular protein. Based on T7 expression vectors originally developed by Studier and colleagues (1-3), high-level expression is achieved because the T7 RNA polymerase is more processive than native E. coliRNA polymerase and is dedicated to the transcription of your gene of interest. Protein production is further enhanced in the system by the expression strain BL21 Star™ E. coli, which significantly improves the stability of mRNA transcripts and increases protein expression up to ten-fold.
Simplified, efficient Directional TOPO™ cloning allows you to quickly enter a Champion™ pET Expression vector. These kits feature linearized, topoisomerase I-activated Champion™ pET expression vectors for five-minute directional cloning. Directional TOPO™ Cloning technology facilitates gene expression because:
• A proofreading enzyme is used for PCR, resulting in fewer errors in cloned genes
• Greater than 90% of the clones are in the correct orientation for gene expression, reducing the time spent on colony screening
Seven Champion™ pET Directional TOPO™ Expression Vectors are available (Figure 1 and Table 1): Each vector carries a T7lac promoter for high-level expression. Flexible options for simplifying protein detection, cleaving purification tags, selecting plasmid carrying clones, and/or improving protein yields are available.
With the Champion™ pET Directional TOPO™ vectors you can expect highest-level protein production. Figure 2 shows expression of the lacZ gene in Champion™ pET Directional TOPO™ vectors. Figure 3 demonstrates efficient cleavage using TEV protease of the N-terminal tag of a β-galactosidase fusion protein expressed from pET151/D-TOPO™.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Bacterial or Yeast StrainBL21 Star™(DE3)
Constitutive or Inducible SystemInducible
Expression MechanismCell-Based Expression
Expression SystemE. coli
Inducing AgentIPTG
Product TypeTOPO Expression Kit
Quantity20 reactions
Selection Agent (Eukaryotic)None
VectorpET
Cloning MethodDirectional TOPO
Product LineOne Shot
PromoterT7, lacO
Protein TagHis Tag (6x), V5 Epitope Tag
Unit SizeEach
Contents & Storage
Each Champion™ pET Directional TOPO™ Expression Kit is provided as a complete expression system. The Directional TOPO™ Expression box contains 200 ng of linearized, topoisomerase I-activated Champion™ pET vector; sterile water; dNTPs; 10X PCR Buffer; salt solution; control template and primers; primers for sequencing or PCR screening; and an expression control. Store at -20°C. The One Shot™ TOP10 box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. The One Shot™ BL21 Star™(DE3) box contains twenty-one 50-μl aliquots of chemically competent E. coli, S.O.C. medium, and a control plasmid. Store at -80°C. Kits with Lumio™ Technology include 20 μl of the Lumio™ Detection Reagent. Store at -20°C. Guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
My gene of interest is toxic to bacterial cells. Are there any precautions you can suggest?
Several precautions may be taken to prevent problems resulting from basal level expression of a toxic gene of interest. These methods all assume that the T7-based or Champion-based expression plasmid has been correctly designed and created.
- Propagate and maintain your expression plasmid in a strain that does not contain T7 RNA polymerase (i.e., DH5α).
- If using BL21 (DE3) cells, try growing cells at room temperature rather than 37 degrees C for 24-48 hr.
- Perform a fresh transformation using a tightly regulated E. coli strain, such as BL21-AI cells.
- After following the transformation protocol, plate the transformation reaction on LB plates containing 100 µg/mL ampicillin and 0.1% glucose. The presence of glucose represses basal expression of T7 RNA polymerase.
- Following transformation of BL21-AI cells, pick 3 or 4 transformants and inoculate directly into fresh LB medium containing 100 µg/mL ampicillin or 50 µg/mL carbenicillin (and 0.1% glucose, if desired). When the culture reaches an OD600 of 0.4, induce expression of the recombinant protein by adding L-arabinose to a final concentration of 0.2%.
- When performing expression experiments, supplement the growth medium with 0.1% glucose in addition to 0.2% arabinose.
- Try a regulated bacterial expression system such as our pBAD system.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm trying to express my protein using a bacterial expression system. How do I know if I'm seeing degradation of my protein or if what I’m seeing is codon usage bias?
Typically, if you see 1-2 dominant bands, translation stopped prematurely due to codon usage bias. With degradation, you usually see a ladder of bands. With degradation, you can try using a protease inhibitor and add it to the lysis buffer to help prevent degradation. If degradation is the issue, a time point experiment can be done to determine the best time to harvest the cells.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm trying to express my protein using a bacterial expression system and am getting inclusion bodies. What should I do?
If you are having a solubility issue, try to decrease the temperature or decrease the amount of IPTG used for induction. You can also try a different, more stringent cell strain for expression. Adding 1% glucose to the bacterial culture medium during expression can also help.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm getting low protein yield from my bacterial expression system. What can I do to improve this?
- Inoculate from fresh bacterial cultures, since higher protein yields are generally obtained from a fresh bacterial colony.
- Check the codon usage in the recombinant protein sequence for infrequently used codons. Replacing the rare codons with more commonly used codons can significantly increase expression levels. For example, the arginine codons AGG and AGA are used infrequently by E. coli, so the level of tRNAs for these codons is low.
- Add protease inhibitors, such as PMSF, to buffers during protein purification. Use freshly made PMSF, since PMSF loses effectiveness within 30 min of dilution into an aqueous solution.
- If you are using ampicillin for selection in your expression experiments, you may be experiencing plasmid instability due to the absence of selective conditions. This occurs as the ampicillin is destroyed by β-lactamase or hydrolyzed under the acidic media conditions generated by bacterial metabolism. You may want to substitute carbenicillin for ampicillin in your transformation and expression experiments.
- The recombinant protein may be toxic to bacterial cells. Try a tighter regulation system for competent cell expression such as BL21-AI. You may also consider trying a different expression system such as the pBAD system.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
My cells are growing very slowly, and I'm not getting any protein expression from my baterial expression system. What can I do to fix this?
This typically occurs when your gene of interest is toxic. Try using a tighter regulation system, such as BL21 (DE3) (pLysS) or BL21 (DE3) (pLysE), or BL21(AI).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.