This product will be discontinued by May 1, 2016. We currently do not offer a direct replacement. For additional assistance, please contact our customer care team.
PCR Optimized Buffer C
Catalog number: K122002C
Invitrogen™  Related applications: PCR
Error loading your content!
  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
PCR Optimized Buffer C is one of ten reaction buffers included in the PCR Optimizer™ Kit, made available here for purchase separately.
PCR Optimizer Kit
The PCR Optimizer Kit is designed to give the researcher a simple way to rapidly determine optimal buffer conditions for all primer-template combinations. Varying the pH level, magnesium concentration, and DMSO content in PCR alters the specificity of priming, which eliminates background products. This enhances the performance of all primer-template combinations, even those with limited homology. Once optimal PCR conditions have been identified, individual PCR Optimizer™ Buffers and dNTPs can be purchased separately. The Optimized™ Buffer Kits are available in convenient 200- and 800-reaction sizes to ensure perfect PCR every time.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Polymerase: Taq DNA Polymerase
Product Size: 200 reactions
Shipping Condition: Dry Ice
Documents
Manuals & protocols
Recommended products