This product is only available through your local Thermo Scientific supplier. Learn where to buy >
EpiJET 5-hmC Enrichment Kit
Catalog number: K1491BID
Thermo Scientific™  Related applications: Methylation Analysis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Thermo Scientific EpiJET 5-hmC Enrichment Kit enables the enrichment of 5-hmC containing DNA over unmodified DNA and 5-mC DNA. The kit takes advantage of the 5-hmC Modifying Enzyme, which is formulated for highly specific and efficient modification of 5-hmC present in CpG dinucleotides of DNA and is deficient for any activity over unmodified or methylated cytosines. The enzyme adds a short linker molecule to 5-hmC, to which biotin is conjugated in the following chemical modification step. The resulting modified DNA is separated from unmodified DNA using streptavidin-coated magnetic beads.
More than 50-fold enrichment of the 5-hmC modified DNA over unmodified and 5-mC containing DNA and at least 20% recovery of the input 5-hmC DNA can be achieved. For performance evaluation of the kit, specific DNA controls (unmodified and 5-hmC DNA) are included.
Highlights
Specific—no activity over unmodified or methylated cytosines
Versatile—efficient enrichment of single and multiple 5-hmC targets
Robust—consistent yield and enrichment
Fast—procedure completed in 3 hours
Pure—enriched DNA suitable for downstream applications
Applications
• PCR
• qPCR
• Sequencing
• Microarrays
• NGS
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line: EpiJET
Product Size: 25 reactions
Contents & storage
Box 1/2 includes:
• 5-hmC Modifying Enzyme
• 4X Enzyme Reaction Buffer
• 10X Biotin Conjugation Buffer
• Biotin Reagent
• Primer Mix for Control DNA (unmodified), 5 µM
• Primer Mix for 5-hmC Control DNA, 5 µM
• 5-hmC Control DNA, 10 ng/µL
• Control DNA (unmodified), 0.5 µg/µL
Box 2/2 includes
• Streptavidin Magnetic Beads
• Streptavidin Binding Buffer
• Streptavidin Wash Buffer 1
• Streptavidin Wash Buffer 2
• Water, nuclease-free
• Column Binding Buffer
• Column Wash Buffer (concentrated)
• Elution Buffer
• DNA Purification Micro Columns and Collection Tubes
Documents
Manuals & protocols