Maxima H Minus First Strand cDNA Synthesis Kit
Thermo Scientific™

Maxima H Minus First Strand cDNA Synthesis Kit

Thermo Scientific Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesNo. of Reactions
K1681Kit with dsDNase20 Reactions
K1651Kit only20 Reactions
K1652Kit only100 Reactions
K1682Kit with dsDNase100 Reactions
Catalog number K1681
Price (USD)
-
Includes:
Kit with dsDNase
No. of Reactions:
20 Reactions
Request bulk or custom format
Thermo Scientific Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA. The kit uses the Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of MMLV RT. The enzyme features the highest thermostability among the derivatives of MMLV RT and lacks RNase H activity.

This kit allows synthesis of long cDNAs up to 20 kb at elevated temperatures (up to 65°C), superseding other systems' abilities to produce full-length cDNA. Due to increased synthesis rates the reaction can be completed in 30 minutes.

For reverse transcription the kit uses Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of MMLV RT.

The Maxima H Minus First Strand cDNA Synthesis Kit with dsDNase provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. The kit contains a novel double-strand-specific DNase (dsDNase) engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific dsDNase activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved, and dsDNase treated RNA can be directly added to reverse transcription.

Features of the Maxima H Minus First Strand cDNA Synthesis Kit include:

  • Increased reaction temperatures—the first strand of cDNA can be synthesized within the 42–65°C temperature range
  • High yields of full-length first strand cDNA—with RNA templates up to 20 kb
  • Flexible priming—oligo(dT)18, random hexamer or gene-specific primers
  • Integrated genomic DNA removal step with dsDNase kit

Applications

  • Full-length cDNA synthesis for long transcripts and ORFs
  • Reverse transcription at elevated temperatures to reduce effects of secondary structure
  • cDNA cloning and construct generation
  • RNA-seq & single cell RNA-seq library prep
  • Isoform and transcript characterization
  • Template-switching-based full-length RNA-seq

Additional information about reaction components

  • Maxima H Minus Enzyme Mix contains Maxima H Minus Reverse Transcriptase and RiboLock RNase Inhibitor. RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.
  • Oligo(dT)18 and random hexamer primers are supplied with the kit. Random hexamer primers bind non-specifically and are used to synthesize cDNA from all RNAs in a total RNA population. The oligo(dT)18 primer selectively anneals to the 3'-end of poly(A) RNA, synthesizing cDNA only from poly(A) tailed mRNA. Gene-specific primers may also be used with the kit to prime synthesis from a specified sequence.
  • 10 mM dNTP Mix is a premixed aqueous solution of dATP, dTTP, dCTP, and dGTP.
  • Nuclease-free water is provided for reaction set-up and dilution of sample DNA. The absence of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
IncludesKit with dsDNase
No. of Reactions20 Reactions
Optimal Reaction Temperature50°C to 55°C
QuantityEach
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseMaxima H Minus
Ribonuclease H ActivityReduced
Shipping ConditionDry Ice
Size (Final Product)Up to 20 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)RT-PCR
GC-Rich PCR PerformanceHigh
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• Maxima H Minus Enzyme Mix
• dsDNase
• 10X dsDNase Buffer
• Oligo(dT)18 and Random hexamer primers
• 5X RT Buffer
• dNTP Mix
• Nuclease-free water

Store at –20°C.

Frequently asked questions (FAQs)

How can I inactivate Thermo Scientific dsDNase?

dsDNase can be inactivated by incubating the sample at 55°C for 5 min in the presence of 10 mM DTT.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.

Does dsDNase cleave ssDNA?

No, dsDNase cleaves only double-stranded DNA. However, if ssDNA forms double-stranded structures, it will act as a target for dsDNase. Because of this, long and complex ssDNA may be partially degraded. Therefore we recommend additional dsDNase inactivation step for RT-PCR amplification of targets 3 kb or longer. The inactivation step should be performed by 5 min incubation at 55 degrees C in the presence of 10 mM DTT.

Can dsDNase be added directly to a reverse transcription reaction to remove genomic DNA?

No. RT reaction composition inhibits dsDNase activity, and genomic DNA removal may be incomplete.

When using the Maxima or Maxima H Minus First Strand cDNA Synthesis kits with dsDNase, can the genomic DNA elimination step be omitted?

Yes. dsDNase buffer is not needed for subsequent reverse transcription reaction. If RNA sample is not contaminated with gDNA, dsDNase treatment may be omitted.