Upgrade to Maxima™ H Minus First Strand cDNA Synthesis Kit for better performance. List price reduced by 40%.
RevertAid RT Reverse Transcription Kit
Thermo Scientific™

RevertAid RT Reverse Transcription Kit

The Thermo Scientific RevertAid RT Kit is a complete system for efficient synthesis of first strand cDNA from mRNA orRead more
Have Questions?
Catalog NumberQuantity
K1691500 reactions
Catalog number K1691
Price (USD)
776.65
Online exclusive
914.00
Save 137.35
Each
Quantity:
500 reactions
Request bulk or custom format
Price (USD)
776.65
Online exclusive
914.00
Save 137.35
Each

The Thermo Scientific RevertAid RT Kit is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates. The kit uses RevertAid Reverse Transcriptase (RT), which has lower RNase H activity compared to AMV reverse transcriptase. The enzyme maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. The recombinant RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA from degradation at temperatures up to 55°C.

First strand cDNA synthesized with this system can be directly used as a template in PCR or real-time PCR. It is also ideal as a first step for second strand cDNA synthesis or linear RNA amplification. Radioactively and non-radioactively labeled nucleotides can be incorporated into first strand cDNA for use as a probe in hybridization experiments, including microarrays.

Features of the RevertAid RT Kit include:
• Full-length first strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit includes all the components for RT reactions

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
No. of Reactions500 Reactions
Optimal Reaction Temperature42°C
Quantity500 reactions
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseRevertAid
Ribonuclease H ActivityYes
Shipping ConditionDry Ice
Size (Final Product)Up to 13 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)Real Time PCR (qPCR)
Reaction Speed60 min.
Unit SizeEach
Contents & Storage

• RevertAid Reverse Transcriptase
• Ribolock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Random Hexamer Primer
• Nuclease-free water

Store at –20°C.


Frequently asked questions (FAQs)

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.