Product Image
Invitrogen™

Pichia Expression Kit, original kit

The Original Pichia Expression Kit is designed for high-level expression of recombinant protein in the yeast Pichia pastoris.
Have Questions?
Catalog NumberQuantity
K1710011 Kit
Catalog number K171001
Price (MXN)
-
Quantity:
1 Kit

The Original Pichia Expression Kit is designed for high-level expression of recombinant protein in the yeast Pichia pastoris. The vectors carry the HIS4 gene for selection of transformants on histidine-deficient medium. The Original Pichia Expression Kit provides all of the tools and reagents needed to express a gene of interest from the AOX1 promoter.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Bacterial or Yeast StrainGS115, KM71
Cloning MethodRestriction Enzyme⁄ MCS
Expression MechanismCell-Based Expression
Expression SystemYeast
For Use With (Application)Protein Expression
Product TypeExpression Kit
Protein TagUntagged
Quantity1 Kit
Cell TypeYeast Cells
FormatKit
PromoterAOX1
SpeciesP. pastoris
VectorpPIC
Unit SizeEach
Contents & Storage
The Original Pichia Expression Kit includes 10 μg each of pHIL-D2 and pPIC3.5 vectors for intracellular expression and pHIL-S1 and pPIC9 vectors for secreted expression, Pichia pastoris strains GS115 (his4), KM71(arg4 his4 aox1::ARG4), control strain GS115 Albumin, control strain GS115 β-gal, media (YP and YNB), Spheroplast Transformation Kit, and 2 μg each 5' AOX1, 3' AOX1, and α-factor sequencing primers.

Store vectors and primers at -20°C. Store strains at room temperature.
All components are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

When selecting for blasticidin-resistant transformants in the X-33 strain using pPIC6/pPIC6α vectors, why do I get large and small colonies on YPD plates containing 300 µg/ml blasticidin?

Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.

When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My transformation is not working. Do you have any suggestions?

Here are some suggestinos:

- Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
- If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
- Use more DNA.
- Use freshly made competent cells.
- If the LiCl transformation method is being used, try boiling the carrier DNA.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My spheroplasting of Pichia worked twice, but hasn't worked since. The OD of the culture simply does not drop.

Here are some things to consider:

- If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
- Do not use old cells and make sure that they are in log phase of growth.
- Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
- Make the PEG solution fresh each time and check the pH.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

What are the different kinds of media used for culturing Pichia pastoris and S. cerevisiae?

Following are the rich and minimal media used for culturing Pichia pastoris and S. cerevisiae:

Rich Media:
S. cerevisiae and Pichia pastoris
YPD (YEPD): yeast extract, peptone, and dextrose
YPDS: yeast extract, peptone, dextrose, and sorbitol

Pichia pastoris only
BMGY: buffered glycerol-complex medium
BMMY: buffered methanol-complex medium

Minimal Media (also known as drop-out media):
S. cerevisiae
SC (SD): Synthetic complete (YNB, dextrose (or raffinose or galactose), and amino acids)

Pichia pastoris
MGY: minimal glycerol medium
MD: minimal dextrose
MM: minimal methanol
BMGH: buffered minimal glycerol
BMMH: buffered minimal methanol

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Is there a recommended protocol for fermentation using constitutive expression vectors such as pGAPZ?

Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:

1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
4) Yeast extract and peptone may be added to the medium for protein stability.

One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Citations & References (68)

Citations & References
Abstract
Expression and functional characterization of the mammalian intestinal peptide transporter PepT1 in the methylotropic yeast Pichia pastoris.
Authors:Doring F, Theis S, Daniel H
Journal:Biochem Biophys Res Commun
PubMed ID:9126331
The methylotrophic yeast Pichia pastoris was used for heterologous expression of the rabbit intestinal peptide transporter PepT1 and its functional characterization. PepT1 mediates the electrogenic transmembrane transport of di- and tripeptides and peptido-mimetics such as beta-lactam antibiotics and ACE-inhibitors. Functional expression of PepT1 was determined in different recombinant clones by ... More
Recombinant proteinase 3 (Wegener's antigen) expressed in Pichia pastoris is functionally active and is recognized by patient sera.
Authors:Harmsen MC, Heeringa P, van der Geld YM, Huitema MG, Klimp A, Tiran A, Kallenberg CG
Journal:Clin Exp Immunol
PubMed ID:9367410
'The open reading frame of human proteinase 3 (PR3) without the prepro- peptide was cloned and expressed in Escherichia coli (rcPR3) and in Pichia pastoris (rpPR3). The 6-histidine tagged rpPR3 was efficiently secreted into culture supernatant from which it could be purified by immobilized metal chelate chromatography. Purified rpPR3 migrated ... More
Autocatalytic processing of recombinant human procathepsin L. Contribution of both intermolecular and unimolecular events in the processing of procathepsin L in vitro.
Authors:Menard R, Carmona E, Takebe S, Dufour E, Plouffe C, Mason P, Mort JS
Journal:J Biol Chem
PubMed ID:9468501
'The autocatalytic processing of procathepsin L was investigated in vitro using purified recombinant proenzyme expressed in Pichia pastoris. Pure intermolecular processing was studied by incubating the mutant procathepsin L (C25S), which cannot autoactivate with a small amount of mature active cathepsin L. The results clearly establish that, contrary to recent ... More
Lethal effect of recombinant human Fas ligand in mice pretreated with Propionibacterium acnes.
Authors:Tanaka M, Suda T, Yatomi T, Nakamura N, Nagata S
Journal:J Immunol
PubMed ID:9036978
'Fas ligand (FasL) is a type II membrane protein. Binding of FasL to its receptor, Fas, induces apoptosis. Matrix metalloproteinase cleaves the membrane-bound human FasL to yield the active soluble form. Here, we have produced a large amount of human soluble rFasL using the yeast, Pichia pastoris. The purified rFasL ... More
Role of the cysteine residues in the alpha1,2-mannosidase involved in N- glycan biosynthesis in Saccharomyces cerevisiae. The conserved Cys340 and Cys385 residues form an essential disulfide bond.
Authors:Lipari F, Herscovics A
Journal:J Biol Chem
PubMed ID:8910350
'The Saccharomyces cerevisiae alpha1,2-mannosidase, which removes one specific mannose residue from Man9GlcNAc2 to form Man8GlcNAc2, is a member of a family of alpha1,2-mannosidases with similar amino acid sequences. The yeast alpha1,2-mannosidase contains five cysteine residues, three of which are conserved. Recombinant yeast alpha1, 2- mannosidase, produced as the soluble catalytic ... More