TA Cloning&trade; Kit, Dual Promoter, with pCR&trade;II Vector and One Shot&trade; TOP10F' Chemically Competent <i>E. coli</i>
Invitrogen™

TA Cloning™ Kit, Dual Promoter, with pCR™II Vector and One Shot™ TOP10F' Chemically Competent E. coli

The TA Cloning™ Kit Dual Promoter with pCR™II vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplifiedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K20604040 Reactions
K20600120 Reactions
Catalog number K206040
Price (USD)
1,464.00
Each
No. of Reactions:
40 Reactions
Price (USD)
1,464.00
Each
The TA Cloning™ Kit Dual Promoter with pCR™II vector provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into a plasmid vector. The T7 and Sp6 promoters of the pCR™II vector allow in vitro transcription of the insert to produce sense or anti-sense products. The TA Cloning Kit Dual Promoter uses the pCR™II cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

Features of the TA Cloning™ Kit Dual Promoter:
• Fast & convenient—15-minute, room-temperature ligation
• Efficient—blue/white screening and >80% clones with correct insert
• Flexible—choice of kanamycin or ampicillin resistance for flexible antibiotic selection
• Hassle-free—eliminates any enzymatic modifications of the PCR product
• Streamlined—does not require the use of PCR primers that contain restriction sites

The pCR™II vector provides:
• 3'-T overhangs for direct ligation of Taq-amplified PCR products
• T7 and Sp6 promoters for in vitro RNA transcription and sequencing
• Versatile polylinker with flanking EcoR I sites for easy excision of inserts
• M13 forward and reverse primer sites for sequencing

How TA Cloning™ Works
Taq polymerase has a non-template-dependent activity that adds a single deoxyadenosine (A) to the 3' ends of PCR products. The linearized vector supplied in this kit has single 3' deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.

Kit Configurations
The TA Cloning™ Kit is offered in a variety of configurations: with One Shot™ INVF' Chemically Competent E. coli (K2050-01 and K2050-40), One Shot™ TOP10F' Chemically Competent E. coli (K2060-01 and K2060-40), and without competent cells (K2750-20 and K2750-40) in 20- and 40-reaction kit sizes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial ContentTOP10
Bacterial or Yeast StrainTOP10F ́
Cell TypeChemically Competent E. coli
Cloning MethodTA Cloning
For Use With (Application)PCR Cloning
IncludesLinearized pCRII vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot TOP10F' chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
No. of Reactions40 Reactions
Product LineOne Shot
Product TypeCloning Kit
PromoterT7, SP6
Quantity40 rxns
VectorpCRII
FormatKit
Unit SizeEach
Contents & Storage
This kit contains linearized pCR™II vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

Can I use TOP10F' competent cells for transformation of my TOPO vector that contains the ccdB gene?

Strains that contain an F plasmid, such as TOP10F', are not recommended for transformation and selection of recombinant clones in any TOPO vector containing the ccdB gene. While the F plasmid does encode the CcdA protein, which acts as an inhibitor of the CcdB gyrase-toxin protein, the half-life of the CcdA protein is shorter than that of the CcdB protein. Overexpression of the CcdB protein causes cell death when its action is not prevented by sufficient CcdA.

Have you compared in vitro transcription levels between SP6 and T7 promoters in your pCRII vectors?

No, we have not done in-house comparisons of transcription levels. It is widely known though that T7 polymerase produces more RNA than SP6 (on the order of 10-fold higher).

Citations & References (10)

Citations & References
Abstract
Semaphorin III can function as a selective chemorepellent to pattern sensory projections in the spinal cord.
Authors:Messersmith EK, Leonardo ED, Shatz CJ, Tessier-Lavigne M, Goodman CS, Kolodkin AL
Journal:Neuron
PubMed ID:7748562
'Distinct classes of primary sensory neurons in dorsal root ganglia subserve different sensory modalities, terminate in different dorsoventral locations in the spinal cord, and display different neurotrophin response profiles. Large diameter muscle afferents that terminate in the ventral spinal cord are NT-3 responsive, whereas small diameter afferents subserving pain and ... More
A nod factor binding lectin with apyrase activity from legume roots.
Authors:Etzler ME, Kalsi G, Ewing NN, Roberts NJ, Day RB, Murphy JB
Journal:Proc Natl Acad Sci U S A
PubMed ID:10318974
'A lectin isolated from the roots of the legume, Dolichos biflorus, binds to Nod factors produced by rhizobial strains that nodulate this plant and has a deduced amino acid sequence with no significant homology to any lectin reported to date. This lectin also is an enzyme that catalyzes the hydrolysis ... More
timrit Lengthens circadian period in a temperature-dependent manner through suppression of PERIOD protein cycling and nuclear localization.
Authors:Matsumoto A, Tomioka K, Chiba Y, Tanimura T
Journal:Mol Cell Biol
PubMed ID:10330175
A fundamental feature of circadian clocks is temperature compensation of period. The free-running period of ritsu (timrit) (a novel allele of timeless [tim]) mutants is drastically lengthened in a temperature- dependent manner. PER and TIM protein levels become lower in timrit mutants as temperature becomes higher. This mutation reduces per ... More
Kainate receptor subunits expressed in single cultured hippocampal neurons: molecular and functional variants by RNA editing.
Authors:Ruano D, Lambolez B, Rossier J, Paternain AV, Lerma J
Journal:Neuron
PubMed ID:7748549
To determine the kainate receptor subunits that are found in native kainate receptors, we have applied a multiplex PCR of cDNAs reverse transcribed from mRNA harvested from single cultured hippocampal neurons after electrophysiological recording. We found that all the cells showing rapidly desensitizing currents in response to kainate express the ... More
Genetic heterogeneity of hypervariable region 1 of the hepatitis C virus (HCV) genome and sensitivity of HCV to alpha interferon therapy
Authors:Sandres K, Dubois M, Pasquier C, Payen JL, Alric L, Duffaut M, Vinel JP, Pascal JP, Puel J, Izopet J
Journal:J Virol
PubMed ID:10623727
Hepatitis C virus (HCV) populations persist in vivo as a mixture of heterogeneous viruses called quasispecies. The relationship between the genetic heterogeneity of these variants and their responses to antiviral treatment remains to be elucidated. We have studied 26 virus strains to determine the influence of hypervariable region 1 (HVR-1) ... More