We have an endotoxin-free (< 0.1 EU/µg) version of this kit available here »
PureLink&trade; HiPure Expi Plasmid Megaprep Kit
Invitrogen™
PureLink™ HiPure Expi Plasmid Megaprep Kit
The PureLink™ HiPure Expi Plasmid Megaprep Kit enables isolation of the highest levels of transfection-grade plasmid DNA from a singleRead more
Have Questions?
Catalog NumberQuantity
K210008XP4 Preps
Catalog number K210008XP
Price (USD)
414.65
Online exclusive
436.00
Save 21.35
Each
Quantity:
4 Preps
Price (USD)
414.65
Online exclusive
436.00
Save 21.35
Each
The PureLink™ HiPure Expi Plasmid Megaprep Kit enables isolation of the highest levels of transfection-grade plasmid DNA from a single E.coli mega prep using an enhanced anion exchange resin. The PureLink™ HiPure Plasmid Megaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times. The PureLink™ HiPure Expi Plasmid Megaprep Kit typically isolates 4 mg of high quality, ultrapure plasmid DNA with inherently low endotoxin levels from 500 mL of bacterial culture.
Advantages of the PureLink™ HiPure Expi Plasmid Megaprep Purification Kit include:
• High yield—isolate over 5 mg of high quality plasmid DNA from a single purification using 1 L of bacterial culture volume
• Purity—inherently low endotoxin levels (typically 0.1–1.0 EU/μg) and A260/280 >1.8, making it ideal for mammalian cell transfection
• Simple and super-fast protocol—with vacuum assisted protocol, typical plasmid isolation takes as little as 90 minutes
HiPure Expi anion exchange chromatography for plasmid purification
PureLink™ HiPure Expi plasmid purification kits use a novel, proprietary anion-exchange resin to enable the purification of the highest levels of transfection-grade plasmid DNA. The enhanced resin combines large capacity with fast flow rate, high resolution, and efficient endotoxin removal. Plasmid preparations are typically completed in less than 2 hours with purity equal to two CsCl gradients. PureLink™ HiPure Expi plasmid purification kits enable the quickest process for obtaining the highest amount of transfection-grade plasmid DNA.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput CompatibilityManual Protocols
No. of Reactions4 Preps
Prep Scale4 mg plasmid DNA
Product LinePureLink
Product TypePlasmid MegaPrep Kit
Quantity4 Preps
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
TargetPlasmid DNA
Test Time2 hr.
FormatKit
Isolation TechnologyAnion Exchange Resin
Unit SizeEach
Contents & Storage
• Equilibration Buffer
• Resuspension Buffer
• Lysis Buffer
• Precipitation Buffer
• RNase A
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi HiPure Mega Filtration Column
• Expi HiPure Mega DNA Binding Column
Store all components at room temperature upon receipt.
Citations & References (1)
Citations & References
Abstract
Structural basis for ADAM17 activation by the iRhom1 pseudoprotease.
Authors:Ungvary JN,Maciag JJ,Alnajjar HF,Dávila EM,Slone CE,Thomas JE,Rich MF,Carazo SA,Ifergan I,Perez-Aguilar JM,Blobel CP,Seegar TCM
Journal:Cell reports
PubMed ID:42024498
The a disintegrin and metalloproteinase (ADAM)-17 releases pro-inflammatory cytokines, including tumor necrosis factor-α, and several epidermal growth factor receptor ligands from cells. ADAM17 is post-translationally regulated by the inactive rhomboid pseudoproteases iRhom1 and iRhom2, and dysregulation of this signaling axis contributes to diseases ranging from chronic inflammation to cancer. Here, ... More