PureLink™ HiPure Expi Plasmid Purification Kits
Invitrogen™

PureLink™ HiPure Expi Plasmid Purification Kits

The PureLink™ HiPure Plasmid Plasmid Purification Kits also include vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호제품 유형수량
K210009XPPlasmid GigaPrep Kit2 Preps
K210009XP-1Rnase1.5 mL
K210009XP-2Binding Cartridge2 Units
K210009XP-3Filtration Column2 Units
K210009XP-4TE Buffer30 mL
K210009XP-5Resuspension Buffer (R3)250 mL
K210009XP-6Lysis Buffer (L7)250 mL
K210009XP-7Precipitation Buffer (N3)250 mL
K210009XP-8Equilibration Buffer (EQ1)400 mL
K210009XP-9Wash Buffer (W8)3 x 400 mL
K210009XP-10Elution Buffer (E4)400 mL
카탈로그 번호 K210009XP
제품 가격(KRW)
363,000
온라인 행사
Ends: 31-Dec-2026
427,000
할인액 64,000 (15%)
Each
제품 유형:
Plasmid GigaPrep Kit
수량:
2 Preps
제품 가격(KRW)
363,000
온라인 행사
Ends: 31-Dec-2026
427,000
할인액 64,000 (15%)
Each

The PureLink™ HiPure Plasmid Plasmid Purification Kits also include vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.

The PureLink™ HiPure Expi Plasmid Plasmid Purification Kits enable isolation of the highest levels of transfection-grade plasmid DNA from a single E. coli prep using an enhanced anion exchange resin. The PureLink™ HiPure Plasmid Plasmid Purification Kits also include vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times. The PureLink™ HiPure Expi Plasmid Plasmid Purification Kits typically isolate 12 mg of high quality, ultrapure plasmid DNA with inherently low endotoxin levels from 2.5 L of bacterial culture.

Advantages of the PureLink™ HiPure Expi Plasmid Plasmid Purification Kits include:

  • High yield—isolate up to 14 mg of high quality plasmid DNA from a single purification using 5 L of bacterial culture volume
  • Purity—inherently low endotoxin levels (typically 0.1–1.0 EU/μg) and A260/280 >1.8, making it ideal for mammalian cell transfection
  • Simple and super-fast protocol—with vacuum assisted protocol, typical plasmid isolation in as little as 90 minutes

HiPure Expi anion exchange chromatography for plasmid purification

PureLink™ HiPure Expi plasmid purification kits use a novel, proprietary anion-exchange resin to enable the purification of the highest levels of transfection-grade plasmid DNA. The enhanced resin combines large capacity with fast flow rate, high resolution, and efficient endotoxin removal. Plasmid preparations are typically completed in less than 2 hours with purity equal to two CsCl gradients. PureLink™ HiPure Expi plasmid purification kits enable the quickest process for obtaining the highest amount of transfection-grade plasmid DNA.

For Research Use Only.
사양
용도(애플리케이션)Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
고처리량 호환성Manual Protocols
프렙 스케일12 mg plasmid DNA
제품라인PureLink
제품 유형Plasmid GigaPrep Kit
수량2 Preps
샘플 종류Bacterial Culture
배송 조건Approved for shipment at Room Temperature or on Wet Ice
타겟Plasmid DNA
테스트 시간2 hr.
형식Kit
Isolation TechnologyAnion Exchange Resin
Unit SizeEach
구성 및 보관
• Equilibration Buffer
• Resuspension Buffer
• Lysis Buffer
• Precipitation Buffer
• RNase A
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi HiPure Giga Filtration Column
• Expi HiPure Giga DNA Binding Column

Store all components at room temperature upon receipt.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Structural basis for ADAM17 activation by the iRhom1 pseudoprotease.
Authors:Ungvary JN,Maciag JJ,Alnajjar HF,Dávila EM,Slone CE,Thomas JE,Rich MF,Carazo SA,Ifergan I,Perez-Aguilar JM,Blobel CP,Seegar TCM
Journal:Cell reports
PubMed ID:42024498
The a disintegrin and metalloproteinase (ADAM)-17 releases pro-inflammatory cytokines, including tumor necrosis factor-α, and several epidermal growth factor receptor ligands from cells. ADAM17 is post-translationally regulated by the inactive rhomboid pseudoproteases iRhom1 and iRhom2, and dysregulation of this signaling axis contributes to diseases ranging from chronic inflammation to cancer. Here, ... More