PureLink™ Quick Gel Extraction Kit
Invitrogen™

PureLink™ Quick Gel Extraction Kit

The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K21001250 Preps
K210025250 Preps
Catalog number K210012
Price (USD)
147.65
Online exclusive
154.00
Save 6.35
Each
Quantity:
50 Preps
Price (USD)
147.65
Online exclusive
154.00
Save 6.35
Each
The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages. DNA can be extracted and purified from agarose gels with different melting points in ∼30 minutes using PureLink™ silica membrane-based quick gel extraction columns. For your convenience, purification protocols are provided for centrifugation and for vacuum.

Advantages of using PureLink™ Quick Gel Extraction Kit:

  • Purify DNA fragments from TAE and TBE agarose gels of various percentages and melting points
  • Complete the procedure in ∼30 minutes
  • Easily purify DNA fragments from 40 bp to 10 kb from gels
  • Obtain high recovery of DNA fragments
  • Bind and purify up to 15 μg DNA with one column
  • Purify DNA fragments that are high quality and show reliable performance in PCR, restriction enzyme digestion, cloning, and labeling

Note: The PureLink™ Quick Gel Extraction Kit is not designed to purify supercoiled plasmid DNA or genomic DNA from agarose gels. Only linear DNA fragments may be purified from gels using this kit.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume50 μL
Final Product TypeDNA
For Use With (Application)PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Label or DyeEthidium Bromide, SYBR Safe
Quantity50 Preps
Sample TypeAgarose Gels, Gel Samples
Shipping ConditionRoom Temperature
Starting Material Amount≤400 mg
Test Time30 min.
Yield15 μg (Binding capacity)
Isolation TechnologySilica Spin Column
Unit SizeEach
Contents & Storage
PureLink Elution Tubes
For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.

Frequently asked questions (FAQs)

My downstream enzymatic reactions are not working after purifying my DNA with your PureLink Quick Gel Extraction Kit. What should I do?

Here are some suggestions for your experiments:

- Many enzymes, including restriction endonucleases and ligases, are inhibited by small amounts of agarose and by perchlorate contaminants. This is not normally a problem. If it is, however, you can use the DNA without repurification by increasing the amount of enzyme used in the digest or ligation or by increasing the digestion incubation time. Also, you may use less DNA in your digest or ligation with the same total volume and the same amount of restriction enzyme.
- There may have been residual ethanol in the eluted fragment. Be sure to thoroughly centrifuge to remove the wash buffer, discard the wash buffer, and use a fresh tube to collect the eluted DNA. For applications that are very sensitive to ethanol, let the open spin column stand for 15 minutes at room temperature to let any excess ethanol evaporate.
- The washing steps may not be as efficient as they should be. Under these circumstances, there may be trace amounts of perchlorate in the eluate. To avoid this, extend the centrifugation times to 5 minutes and wash 2 times with wash buffer.
- Be sure to perform the optional wash step if you are using higher concentrations of agarose or are adding more than 250 mg to the cartridge. If applications are sensitive to EDTA, elute with water (pH 7.5-8.5), or with 10 mM Tris, pH 8.0 without EDTA.

What types and concentrations of agarose are compatible with the PureLink Quick Gel Extraction Kit (Cat. No. K210012)?

Both regular and low-melting agaroses can be used. When the agarose concentration is above 2%, use 600 µL of solubilization buffer for each 100 mg of gel.

Can I use water to elute my sample using the PureLink Quick Gel Extraction Kit?

Yes, water may be used, but please ensure that the water is clean and the pH of the water is between 7.5 and 8.5.

I'm running an agarose gel with TAE or TBE. Can I use your gel extraction kits?

Yes, both TAE and TBE agarose gels are compatible with our gel extraction kits.

Why did I get multiple bands on post-extraction gel analysis after using the PureLink Quick Gel Extraction Kit (Cat. No. K210012)?

Multiple bands can occur when the DNA is partially denatured on the gel due to high heat generated during electrophoresis or running the gel too fast. This can appear as multiple bands when the eluted DNA is analyzed on a gel. Denaturation can also occur in AT-rich DNA during the 50 degrees C incubation to dissolve the gel slices. If this happens, solubilize the gel at 37 degrees C for 20 to 30 minutes with repeated vortexing.

Citations & References (1)

Citations & References
Abstract
A naturally occurring splice variant of CXCL12/stromal cell-derived factor 1 is a potent human immunodeficiency virus type 1 inhibitor with weak chemotaxis and cell survival activities.
Authors:Altenburg JD, Broxmeyer HE, Jin Q, Cooper S, Basu S, Alkhatib G,
Journal:J Virol
PubMed ID:17507482
CXCL12/stromal cell-derived factor 1 is a member of the CXC family of chemokines that plays an important role in hematopoiesis and signals through CXCR4 and CXCR7. Two splice variants of human CXCL12 (CXCL12alpha and CXCL12beta) induce chemotaxis of CXCR4(+) cells and inhibit X4 infection. Recent studies described four other novel ... More