PureLink™ HiPure BAC Buffer Kit
Invitrogen™
PureLink™ HiPure BAC Buffer Kit
The PureLink™ HiPure BAC Buffer Kit provides additional buffers and reagents for performing BAC (Bacterial Artificial Chromosome) DNA isolation usingRead more
Have Questions?
Catalog NumberQuantity
K210018
also known as K2100-18
1 kit
Catalog number K210018
also known as K2100-18
Price (TWD)
4,700.00
Online offer
Ends: 31-Dec-2026
6,260.00
Save 1,560.00 (25%)
Each
Quantity:
1 kit
Price (TWD)
4,700.00
Online offer
Ends: 31-Dec-2026
6,260.00
Save 1,560.00 (25%)
Each
The PureLink™ HiPure BAC Buffer Kit provides additional buffers and reagents for performing BAC (Bacterial Artificial Chromosome) DNA isolation using the PureLink™ HiPure Plasmid Kits. The PureLink™ HiPure Plasmid Kits utilize a proprietary anion-exchange resin which allows you to purify high-quality BAC DNA from E. coli cultures. BAC DNA requires the use of large bacterial culture volumes to obtain a reasonable DNA yield. The additional buffers and reagents needed for working with larger bacterial cultures are provided in this Buffer Kit. From a 100 ml culture, you can expect to purify ∼40 μg of BAC DNA. The PureLink™ HiPure BAC Buffer Kit provides reagents sufficient for five BAC Maxipreps, or 25 BAC Midipreps, or 100 BAC Minipreps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeBAC DNA
For Use With (Application)Sequencing, Cloning, Transformation, Transfection, In Vitro Transcription
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LinePureLink
Product TypeBAC Buffer Kit
Quantity1 kit
Sample TypeBacterial Culture
Shipping ConditionRoom Temperature
TargetBAC DNA, Plasmid DNA
FormatKit
Isolation TechnologyAnion Exchange Resin
Unit SizeEach
Contents & Storage
The PureLink™ HiPure BAC Buffer Kit includes Resuspension Buffer (R3) (250 mL), Lysis Buffer (L7) (250 mL), Precipitation Buffer (N3) (250 mL), and RNase A (20 mg/mL) (5 mL). All components of the PureLink™ HiPure BAC Buffer Kit are shipped at room temperature. Store all components at room temperature upon receipt.