Invitrogen™

PureLink™ HiPure Plasmid Midi/Maxiprep Precipitators

Catalog number: K210020S
Invitrogen™

PureLink™ HiPure Plasmid Midi/Maxiprep Precipitators

Catalog number: K210020S
Catalog Number
K210020S
also known as K2100-20S
Unit Size
4 Precipitators
Price (USD)
Price: 83.75
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K210020S
also known as K2100-20S
4 Precipitators
Price: 83.75
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The PureLink™ HiPure Precipitator allows fast, simple, and efficient desalting and concentration of plasmid DNA isolated using anion-exchange chromatography. The traditional method for DNA precipitation of anion exchange isolated plasmid DNA involves isopropanol precipitation followed by centrifugation and drying of DNA which is time consuming and labor intensive. The use of the PureLink™ HiPure Precipitator allows plasmid DNA concentration within 5 minutes, eliminates the need for centrifugation, and reduces the risk of losing the DNA pellet during supernatant removal. The recovery is >80% and resulting plasmid DNA is ready to use in the most challenging applications such as transfection.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Isolation Technology
Anion Exchange Resin
Sample Type
Bacterial Culture
Final Product Type
Plasmid DNA
For Use With (Application)
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility
Not High-throughput Compatible (Manual)
Prep Scale
100-200 µg (Medium-Scale) Plasmid DNA
Product Line
PureLink™
Product Type
Plasmid Midi/MaxiPrep Precipitator
Quantity
4 Preps
Shipping Condition
Room Temperature
Target
Plasmid DNA
Test Time
5 min

Contents & Storage

Store at room temperature.

Figures