PureLink™ Quick Gel Extraction and PCR Purification Combo Kit
Invitrogen™

PureLink™ Quick Gel Extraction and PCR Purification Combo Kit

Offers ability to perform both a gel extraction or PCR purification with single kit.
Have Questions?
Catalog NumberQuantity
K22000150 Preps
Catalog number K220001
Price (USD)
157.65
Online exclusive
164.00
Save 6.35
Each
Quantity:
50 Preps
Price (USD)
157.65
Online exclusive
164.00
Save 6.35
Each

PureLink™ Quick Gel Extraction and PCR Purification Combo Kit offers the ability to perform both a gel extraction or a PCR purification a single kit.

The PureLink™ Quick Gel Extraction and PCR Purification Combo Kit is designed to purify DNA fragments from agarose gels. The simple procedure uses a silica-based spin cartridge to purify DNA fragments from 40 bp to 10 kb in < 30 minutes. It also provide all components to perform a rapid and efficient removal of short primers, dNTPs, enzymes, short-failed PCR products, and salts from PCR fragments >100 bp in less than 10 minutes. Isolated DNA using the PureLink™ kit is free of proteins, dye, and agarose, and is ready-to-use for a variety of applications, including automated fluorescent DNA sequencing, manual DNA sequencing, PCR, in vitro transcription, restriction mapping, cloning, and labeling. The kit contains all the buffers from the PureLink™ Quick Gel extraction kit and the PCR binding buffer from PureLink™ PCR Purification kit with Invitrogen branded lidded column.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume50 μL
Final Product TypeDNA
For Use With (Application)PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Label or DyeEthidium Bromide, SYBR Safe
Quantity50 Preps
Sample TypeLinear DNA fragments from: Agarose Gels, PCR Purification, Restriction Enzyme Reactions, PCR Products
ScaleMini
Shipping ConditionRoom Temperature
Starting Material AmountAgarose Gel: ≤400 mg, PCR Product: ≤100 μL, or ≤40 μg dsDNA
Test TimeUp to 30 min.
Yield15 μg (Binding capacity)
Isolation TechnologySilica Spin Column
Unit SizeEach
Contents & Storage
For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.

Frequently asked questions (FAQs)

I'm running an agarose gel with TAE or TBE. Can I use your gel extraction kits?

Yes, both TAE and TBE agarose gels are compatible with our gel extraction kits.

Can the PureLink Quick PCR Clean-Up Kit be used to purify linear/digested or supercoiled plasmid DNA?

Yes, you can purify ANY dsDNA sample with a size range of 100 bp to 12 kb from liquid samples with the PureLink Quick PCR Clean-Up Kit. Supercoiled DNA can be purified as well, but some nicking should be expected.

What is the difference between Purelink PureLink Quick Gel Extraction System and S.N.A.P. Gel Purification Kit?

The PureLink kit can be used with both TAE and TBE gels, while the S.N.A.P. Gel Purification Kit is recommended for TAE gels only.

What is unique about the S.N.A.P. UV-free Gel Purification Kit?

The S.N.A.P. UV-free Gel Purification Kit utilizes crystal violet as a staining reagent, which can be visualized in non-UV light conditions. This can prevent DNA damage that can be caused by the UV light required by normal ethidium bromide staining. DNA fragments recovered using this kit have higher cloning efficiency. In addition, DNA can be visualized under normal light as a thin violet band while the gel is running and excised as soon as bands are sufficiently resolved.