Product Image
Invitrogen™

pCR™8/GW/TOPO™ TA Cloning Kit with One Shot™ Mach1™-T1R E. coli and PureLink™ Kit

The pCR™8/GW/TOPO™ TA Cloning Kit delivers simple 5 minute cloning and access to the Gateway™ recombination cloning vector platform. OurRead more
Have Questions?
Catalog NumberQuantity
K25200220 Preps
Catalog number K252002
Price (USD)
832.00
Each
Quantity:
20 Preps
Price (USD)
832.00
Each
The pCR™8/GW/TOPO™ TA Cloning Kit delivers simple 5 minute cloning and access to the Gateway™ recombination cloning vector platform. Our pCR™8 TOPO™ TA vector contains a minimal MCS, so you sequence more of your insert and less of the vector. And with our patented att sites, the pCR™8 vector is the simplest way to create an Entry Clone, which facilitates easy access via Gateway™ recombination cloning to a variety of expression vectors without tedious sub-cloning, saving you valuable research time.

The pCR™8/GW/TOPO™ TA Cloning Kit features:
• Fast growing E. coli that shortens cloning time
• Maximum convenience: includes plasmid miniprep kit
• att sites for rapid recombination into any Gateway™ destination vector
• 3'-T overhangs for direct ligation of Taq-amplified PCR products
• Sequencing primer sites within 55 bp of insertion site ensures you sequence more insert, less vector
• Spectinomycin resistance gene for robust selection in E. coli
• EcoR I sites flanking the PCR product insertion site for easy excision of inserts

This kit includes One Shot™ Mach1™-T1R Chemically Competent E. coli and a PureLink™ Quick Plasmid Miniprep Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodTOPO™ with Gateway™
No. of Reactions20 preps
Product LineOne Shot
Product TypeTA Cloning Kit
Quantity20 Preps
VectorpCR
Unit SizeEach
Contents & Storage
pCR™8/GW/TOPO™ Vector, dNTPs, Salt Solution, Sterile Water, Gateway Forward and Reverse Sequencing Primers, Control Template and PCR Primers; One Shot™ TOP10 Chemically Competent E. coli Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink™ Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes. Store all components of the PureLink™ kit at room temperature. Store the One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

Can I store my competent E. coli in liquid nitrogen?

We do not recommend storing competent E. coli strains in liquid nitrogen as the extreme temperature can be harmful to the cells. Also, the plastic storage vials are not intended to withstand the extreme temperature and may crack or break.

How should I store my competent E. coli?

We recommend storing our competent E. coli strains at -80°C. Storage at warmer temperatures, even for a brief period of time, will significantly decrease transformation efficiency.

Your Gateway-adapted TOPO vectors are supplied with a control template and control primers. Can I obtain the sequence of the control template?

The sequence of the control template is proprietary.

What is the best ratio of insert:vector to use for cloning? Is there an equation to calculate this?

The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio

Does Platinum Taq DNA Polymerase High Fidelity enzyme mix leave 3' A-overhangs on the PCR product for subsequent cloning into a TOPO TA or original TA vector?

Yes, the enzyme mix leaves 3' A-overhangs on a portion of the PCR products. However, the cloning efficiency is greatly decreased compared to that obtained with Taq polymerase alone. It is recommended to add 3' A-overhangs to the product for TA cloning.