Maxima H Minus Double-Stranded cDNA Synthesis Kit
Thermo Scientific™

Maxima H Minus Double-Stranded cDNA Synthesis Kit

Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K256110 Reactions
K256250 Reactions
K2563200 Reactions
Catalog number K2561
Price (USD)
658.65
Online exclusive
708.00
Save 49.35
Each
Quantity:
10 Reactions
Request bulk or custom format
Price (USD)
658.65
Online exclusive
708.00
Save 49.35
Each
Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA. First- and second-strand cDNA synthesis reactions are performed in the same tube without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains pre-mixed components to reduce the number of pipetting steps necessary to complete the procedure.

Features of the Maxima H Minus Double-Stranded cDNA Synthesis Kit include:
• Efficient synthesis of full-length double-stranded cDNA
• Fast—procedure completed in less than two hours
• Convenient—pre-mixed components
• Complete—includes all primers, controls and residual RNA removal reagents

Applications
• Full-length double-stranded blunt-end cDNA synthesis for cloning
• Double-stranded cDNA library construction

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeDouble Stranded cDNA
Format10 reactions
No. of Reactions10 Reactions
Optimal Reaction Temperature50°-55° C
Product LineMaxima
Product TypecDNA Synthesis Kit
Quantity10 Reactions
Reaction FormatSeparate Components
Reverse TranscriptaseMaxima H Minus
Sample TypeRNA
For Use With (Application)Cloning
Unit SizeEach

Frequently asked questions (FAQs)

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.