What is the largest insert size tested with pZErO vectors?
BAC genomic clones as large as 25 kb have been reported to be cloned into the pZErO vectors.
Search
View additional product information for Zero Background™/Kan Cloning Kit - FAQs (K260001)
1 product FAQs found