MagJET Plant RNA Purification Kit - FAQs
MagJET Plant RNA Purification Kit - FAQs
View additional product information for MagJET Plant RNA Purification Kit - FAQs (K2772)
6 product FAQs found
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Are MagJET Plant Genomic DNA and RNA Kits compatible with polyphenol rich samples?
The kits include recommendations in the protocols for polyphenol rich and woody samples such as branches, twigs, needles, etc.
Are the MagJET Plant Kits compatible with sample disruption using automated homogenizers?
MagJET Plant Kits are compatible with homogenization using liquid nitrogen, as well as automated homogenizers.
Which magnetic rack should I use to follow the manual protocol of MagJET kits?
Thermo Scientific offers MagJET Separation Rack of different formats for 1.5 mL tubes (Cat. Nos. MR01, MR02), 50 mL tubes (Cat. No. MR04), and 96-well plates (Cat. No. MR03). You may use other compatible magnetic racks available in the market as well.
Is a sample size available for MagJET kits?
No, we do not offer a sample size for MagJET kits.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.