MagJET Plasmid DNA Kit
Thermo Scientific™
MagJET Plasmid DNA Kit
Thermo Scientific™ MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
K279196 Preps
K27924 x 96 Preps
카탈로그 번호 K2791
제품 가격(KRW)
221,000
온라인 행사
Ends: 31-Dec-2026
259,000
할인액 38,000
Each
수량:
96 Preps
제품 가격(KRW)
221,000
온라인 행사
Ends: 31-Dec-2026
259,000
할인액 38,000
Each
Thermo Scientific™ MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET Magnetic Beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput users.
The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors. Purified plasmid DNA can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.
Highlights
  • High yields – up to 5 μg of plasmid DNA from 1 mL of bacterial culture
  • Pure – isolated DNA is immediately ready-to-use in downstream application such as sequencing, transcription, digestion with restriction enzymes or transfection of robust cell lines
  • Flexible – automation and manual processing protocols available
  • Repeatable results and robust performance
Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
  • FastDigest or conventional restriction digestion
  • Automated fluorescent and radioactive sequencing
  • PCR
  • In vitro transcription
  • Transformation
  • Transfection of robust cell lines
For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
고처리량 호환성High-throughput Compatible, Not High-throughput Compatible (Manual)
프렙 스케일< 100 μg (Small-Scale) Plasmid DNA
제품라인MagJET
제품 유형Plasmid DNA Kit
수량96 Preps
샘플 종류Bacteria
타겟Plasmid DNA
형식Kit
Isolation TechnologyMagnetic Bead
Unit SizeEach
자주 묻는 질문(FAQ)
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?
We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.
Are MagJET and GeneJET plasmid purification kits compatible with EndA+ bacterial strains?
Yes, MagJET and GeneJET Plasmid DNA Purification Kits are suitable for plasmid DNA purification from EndA+ strains. Please refer to the specific kit protocol for details. GeneJET Plasmid Miniprep Kit requires Wash Solution I (Cat. No. R1611, not included in the kit) for EndA+ strains.
Which magnetic rack should I use to follow the manual protocol of MagJET kits?
Thermo Scientific offers MagJET Separation Rack of different formats for 1.5 mL tubes (Cat. Nos. MR01, MR02), 50 mL tubes (Cat. No. MR04), and 96-well plates (Cat. No. MR03). You may use other compatible magnetic racks available in the market as well.
Is a sample size available for MagJET kits?
No, we do not offer a sample size for MagJET kits.
Are GeneJET spin columns or MagJET magnetic beads available separately?
No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.