PureLink™ PCR Micro Kit, 50 Preps - FAQs
PureLink™ PCR Micro Kit, 50 Preps - FAQs
View additional product information for PureLink™ PCR Micro Kit - FAQs (K310250, K310050)
2 product FAQs found
What are the size limitations for the PureLink PCR Cleanup Micro Kit?
The size cut-off is 100 bp to 12 kb.
What are the minimum and maximum size ranges for DNA fragment cleanup using the PureLink PCR Clean-Up Kit?
The size range is between 100 bp and 12 kb. Our kit comes with two buffers, Binding Buffer (B2) and Binding Buffer HC (B3). The Binding Buffer HC (B3) eliminates primer-dimers and short failed PCR products that are smaller than 300 bp.