TOPO&trade; TA Cloning&trade; Kit, Dual Promoter, with pCR&trade;II-TOPO&trade; Vector and One Shot&trade; TOP10F' Chemically Competent <i>E. coli</i>
Invitrogen™

TOPO™ TA Cloning™ Kit, Dual Promoter, with pCR™II-TOPO™ Vector and One Shot™ TOP10F' Chemically Competent E. coli

TOPO™ TA Cloning™ Kits are designed for cloning PCR products directly from a PCR reaction in just 5 minutes (1).Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K46500125 Reactions
K46504050 Reactions
Catalog number K465001
Price (CLP)
951.588
Each
Quantity:
25 Reactions
Price (CLP)
951.588
Each
TOPO™ TA Cloning™ Kits are designed for cloning PCR products directly from a PCR reaction in just 5 minutes (1). They use a pCR™-TOPO™ Vector with covalently bound topoisomerase I for fast cloning and recombinants. pCR™-TOPO™ Vectors include:

• 3´-T overhangs for direct ligation of Taq-amplified PCR products
• Choice of T7 (pCR™2.1-TOPO™) or T7 and SP6 (pCR™II-TOPO™) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
• EcoR I sites flanking the PCR product insertion site for easy excision of inserts
• Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
• Easy blue/white colony screening for selection of recombinants

TOPO™ TA Cloning™ Kits are also available in combo kits combined with a PureLink™ Quick Plasmid Miniprep Kit (50 preps) for fast plasmid purification of your TOPO™-cloned inserts for downstream analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainTOP10F´
Cell TypeChemically Competent
Cloning MethodTOPO™-TA
Product LineOne Shot
Product TypeCloning Kit
PromoterT7, SP6
Quantity25 Reactions
VectorTOPO-TA Cloning Vectors
FormatKit
Unit SizeEach
Contents & Storage
Box 1:
• Topoisomerase I-activated pCR™II-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid.

Store at -68 to -85°C.

Frequently asked questions (FAQs)

Can I store my competent E. coli in liquid nitrogen?

We do not recommend storing competent E. coli strains in liquid nitrogen as the extreme temperature can be harmful to the cells. Also, the plastic storage vials are not intended to withstand the extreme temperature and may crack or break.

How should I store my competent E. coli?

We recommend storing our competent E. coli strains at -80°C. Storage at warmer temperatures, even for a brief period of time, will significantly decrease transformation efficiency.

What is the difference between the pCR2.1-TOPO and pCR4-TOPO vectors?

The vector backbones for both of these vectors are very similar. The main difference is that the pCR4-TOPO vector has sequencing primer sites located as close as 33 base pairs from the PCR product insertion site. This minimizes the amount of vector DNA sequence that needs to be read before reaching the sequence of the insert, making the pCR4-TOPO vector very useful for sequencing applications.

What is the difference between the pCR2.1-TOPO and pCRII-TOPO vectors?

The vector backbones for both of these vectors are very similar. The main difference is that the pCRII-TOPO vector is a dual promoter vector, containing the SP6 and T7 promoters for in vitro transcription/sequencing, whereas the pCR2.1-TOPO vector contains only the T7 promoter for in vitro transcription/sequencing. Both vectors contain the M13 Forward and Reverse primer sites for sequencing or PCR screening.

Your TOPO cloning kits contain a control template and control primers. Can I obtain the sequence of the control template?

The sequence of the control template is proprietary.

Citations & References (1)

Citations & References
Abstract
Cloning and functional expression of a thyrotropin receptor cDNA from rat fat cells.
Authors:Endo T; Ohta K; Haraguchi K; Onaya T;
Journal:J Biol Chem
PubMed ID:7738021
Thyrotropin receptor (TSH-R) has been thought to be thyroid-specific, but, by Northern blot analysis, we found that rat adipose tissue expressed TSH-R mRNAs in amounts approaching those in the thyroid. To investigate the function of TSH-R from adipose tissue, we screened a rat fat cell lambda gt11 cDNA library for ... More