본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
Product Image
Invitrogen™
NT-GFP Fusion TOPO™ Expression Kit
The GFP Fusion TOPO™ TA Expression Kits are designed to allow fusion of a protein of interest to the Cycle자세히 알아보기
Have Questions?
카탈로그 번호수량
K481001
K4810-01으로도 사용됨
20 reactions
카탈로그 번호 K481001
K4810-01으로도 사용됨
제품 가격(KRW)
1,385,000
온라인 행사
Ends: 31-Dec-2026
1,629,000
할인액 244,000 (15%)
Each
수량:
20 reactions
제품 가격(KRW)
1,385,000
온라인 행사
Ends: 31-Dec-2026
1,629,000
할인액 244,000 (15%)
Each
The GFP Fusion TOPO™ TA Expression Kits are designed to allow fusion of a protein of interest to the Cycle 3 GFP protein. These kits provide topoisomerase-I activated pcDNA3.1/NT-GFP-TOPO™ or pcDNA3.1/CT-GFP-TOPO™ vectors (Figure 1). Taq-amplified DNA fragments are ligated into these vectors in a simple 5-minute reaction right on your bench top. The vectors are designed for high-level expression of transient or stable GFP fusion proteins in a wide range of mammalian cells (Figure 2). Expression is easily detected in living cells using fluorescence. In addition, recombinant proteins expressed from these vectors can be detected on western blots using GFP Antiserum.
For Research Use Only. Not for use in diagnostic procedures.
사양
구성 또는 유도성 시스템Constitutive
배달 유형Transfection
용도(애플리케이션)Reporter Assays
제품 유형TOPO Expression Kit
수량20 reactions
리포터 유전자GFP (Cycle 3)
선택 제제(진핵)Geneticin™ (G-418)
벡터TOPO-TA Vectors
클로닝 방법TOPO-TA
제품라인TOPO
프로모터CMV
단백질 태그GFP (Cycle 3)
Unit SizeEach
구성 및 보관
GFP Fusion TOPO™ Expression Kits contain two boxes. The GFP Fusion TOPO™ TA Cloning box contains linearized and topoisomerase I-activated cloning vector, dNTPs, salt solution control PCR template and primers, forward and reverse primers for sequencing and PCR screening, and a GFP expression control plasmid. Store at -20°C. The One Shot ¤ box contains transformation reagents including single-use, 50-μl aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and supercoiled control plasmid. Store the One Shot™ box at -80°C. Kits are guaranteed stable for 6 months when properly stored.
자주 묻는 질문(FAQ)
Can I use pBlue- or pGlow-TOPO reporter vectors to evaluate promoter function in E. coli?
pBlue-TOPO contains a cryptic prokaryotic promoter upstream of the lacZ reporter gene, due to which E. coli transformants may appear to be light blue when screened on plates containing X-Gal. Hence, we do not recommend using pBlue-TOPO to evaluate promoter function in E. coli. However, pGlow-TOPO can be used for these studies. Note that background expression of beta-galactosidase from pBlue-TOPO does not occur in mammalian cells.
Do you offer a vector that I could use to do some promoter analysis studies?
Yes, we do offer the pBlue-TOPO and pGlow-TOPO vectors that facilitate cloning of the DNA sequence of interest directly upstream of either the b-galactosidase or Cycle 3 GFP gene, respectively.
pBlue-TOPO is ideal for functional analysis of promoters with low transcriptional activity, since assays for beta-galactosidase are easy to perform and are quantitative at very low levels of expression. pGlow-TOPO is ideal for non-invasive analysis of promoter elements within intact, living cells. The fluorescent property of Cycle 3 GFP allows in vivo detection in virtually any cell type or species using microscopy with wild-type GFP filter sets or by fluorescence-activated cell sorting methods.
Can GFP fluorescence be detected in cells that have been stained for beta-galactosidase?
We recommend looking for GFP fluorescence before staining for beta-galactosidase. This is because the beta-galactosidase staining process produces a very high autofluorescence that will interfere with detection of GFP fluorescence.
What filter set should be used to detect Cycle 3 GFP fluorescence? How can I measure Cycle 3 GFP fluorescence using a fluorometer and what model fluorometer should I use?
Cycle 3 GFP fluorescence can be detected using a filter set designed to detect wild-type GFP (since they have the same fluorescence spectra). In-house, we use the XF76 filter set from Omega Optical. For Cycle 3 GFP, excite at 395 nm and read emission at 507 nm. You can also look at the emission spectra and record emissions from 200-800 nm.
Cycle 3 GFP fluorescence can be quantitated with any type of fluorometer with the appropriate filters and cut-off wavelengths. In-house, we have a Hitachi F-2000 Fluorescence Spectrophotometer. Our general protocol using this machine is as follows:
Dilute samples in PBS (although Tris or water would be okay). The amount of lysate to be used will of course depend upon the concentration of GFP. This will have to be determined empirically. The primary consideration is that one needs to be in the linear range of the fluorometer. We have used quantities from 5-50 µL in 1 mL of PBS in a cuvette. If readings are going to be internally compared, the most consistent results will be obtained if the amounts of lysate used are normalized to either the transfection efficiency or the total protein concentration.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What are the recommended filter sets for detection of EmGFP, YFP, CFP, and BFP by fluorescence microscopy?
EmGFP, YFP, CFP, and BFP can be detected using standard FITC filter sets and settings. However, for optimal detection of the fluorescence signal, filter sets optimized for detection within the excitation and emission ranges for each fluorescent protein are recommended. The recommended filter sets are as follows: EmGFP: Omega filter set XF100 YFP: Omega filter set XF1042 Chroma filter set 41028 CFP: Omega filter set XF114 Chroma filter set 31044 BFP: Omega filter set XF10 Chroma filter set 31021 For information on obtaining filter sets, please contact Omega Optical, Inc. (www.omegafilters.com) or Chroma Technology Corporation (www.chroma.com) directly.
인용 및 참조 문헌 (6)
인용 및 참조 문헌
Abstract
Subcellular targeting of RGS9-2 is controlled by multiple molecular determinants on its membrane anchor, R7BP.
Authors:Song JH, Waataja JJ, Martemyanov KA,
Journal:J Biol Chem
PubMed ID:16574655
'RGS9-2, a member of the R7 RGS protein family of neuronal RGS (Regulators of G protein Signaling), is a critical regulator of G protein signaling. In striatal neurons, RGS9-2 is tightly associated with a novel palmitoylated protein - R7BP (R7 family Binding Protein). Here we report that R7BP acts to ... More
A nuclear export signal within the high mobility group domain regulates the nucleocytoplasmic translocation of SOX9 during sexual determination.
Authors: Gasca Stephan; Canizares Joaquin; De Santa Barbara Pascal; Mejean Catherine; Poulat Francis; Berta Philippe; Boizet-Bonhoure Brigitte;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12169669
'In mammals, male sex determination starts when the Y chromosome Sry gene is expressed within the undetermined male gonad. One of the earliest effect of Sry expression is to induce up-regulation of Sox9 gene expression in the developing gonad. SOX9, like SRY, contains a high mobility group domain and is ... More
The Erbin PDZ domain binds with high affinity and specificity to the carboxyl termini of delta-catenin and ARVCF.
Authors: Laura Richard P; Witt Andrea S; Held Heike A; Gerstner Resi; Deshayes Kurt; Koehler Michael F T; Kosik Kenneth S; Sidhu Sachdev S; Lasky Laurence A;
Journal:J Biol Chem
PubMed ID:11821434
'Erbin is a recently described member of the LAP (leucine-rich repeat and PDZ domain) protein family. We used a C-terminally displayed phage peptide library to identify optimal ligands for the Erbin PDZ domain. Phage-selected peptides were type 1 PDZ ligands that bound with high affinity and specificity to the Erbin ... More
Cloning and characterization of AOEB166, a novel mammalian antioxidant enzyme of the peroxiredoxin family.
Authors:Knoops B, Clippe A, Bogard C, Arsalane K, Wattiez R, Hermans C, Duconseille E, Falmagne P, Bernard A
Journal:J Biol Chem
PubMed ID:10521424
Using two-dimensional electrophoresis, we have recently identified in human bronchoalveolar lavage fluid a novel protein, termed B166, with a molecular mass of 17kDa. Here, we report the cloning of human and rat cDNAs encoding B166, which has been renamed AOEB166 for antioxidant enzyme B166. Indeed, the deducedamino acid sequence reveals ... More
Molecular and Functional Interaction of the ATP-binding Cassette Transporter A1 with Fas-associated Death Domain Protein.
Authors: Buechler Christa; Bared Salim Maa; Aslanidis Charalampos; Ritter Mirko; Drobnik Wolfgang; Schmitz Gerd;
Journal:J Biol Chem
PubMed ID:12235128
ATP-binding cassette transporter A1 (ABCA1) is a major regulator of cellular cholesterol and phospholipid homeostasis. Its function has not been fully characterized and may depend on the association with additional proteins. To identify ABCA1-interacting proteins a human liver yeast two-hybrid library was screened with the 144 C-terminal amino acids of ... More