Product Image
Invitrogen™

BLOCK-iT™ HiPerform™ Lentiviral Pol II miR RNAi Expression System with EmGFP

The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits combine the advantages of traditional RNAi vectors (stable expression and theRead more
Have Questions?
Catalog NumberQuantity
K49340020 reactions
Catalog number K493400
Price (USD)
4,340.00
Each
Quantity:
20 reactions
Price (USD)
4,340.00
Each
The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits combine the advantages of traditional RNAi vectors (stable expression and the ability to use viral delivery) with capabilities for tissue-specific expression and multiple target knockdown from the same transcript. The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits and the BLOCK-iT™ Lentiviral Pol II miR RNAi Expression Systems are designed to express artificial miRNAs which are engineered to have 100% homology to your target sequence and will result in target cleavage. Using Invitrogen’s award-winning BLOCK-iT™ RNAi Designer, over 70% of constructs produce more than 70% knockdown. The Invitrogen Pol II expression family of vectors include:

–Strong expression from the CMV immediate early promoter, with the option to use EF-1α, tissue-specific promoters, or other promoters via MultiSite Gateway™ recombination.
–Compatibility with many of Invitrogen’s Gateway™ destination (DEST) vectors for gene expression.

–Ability to control the initiation of the RNAi response with a new BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit w/EmGFP (cat. no. K493900).
–A new destination vector in the BLOCK-iT™ HiPerform™ Lentiviral PolII miR RNAi Expression System with EmGFP. The new vector contains an mRNA stabilizing sequence (WPRE) and a nuclear import sequence (cPPT) which generate up to 5-fold higher virus titers and EmGFP expression levels. Additionally, blasticidin resistance is expressed from the mouse PGK-1 promoter to avoid shut-down after multiple passages.

–Co-cistronic expression of Emerald GFP (EmGFP), resulting in correlation of EmGFP expression with knockdown from your miR RNAi.

–Expression of more than one engineered miR RNAi sequence on the same transcript, allowing the knockdown of multiple genes simultaneously and the generation of synthetic phenotypes

The new BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP provides the ability to regulate RNAi experiments. This kit contains the pT-REx-DEST30 Gateway™ vector which after simple cloning and shuttling techniques, produces a miR RNAi expression vector suitable for inducible knockdown. The pT-REx-DEST30 Gateway™ vector contains the CMV promoter with two copies of the tetracycline operator (tetO2) sequence allowing high-level and regulated expression. This permits the study of loss of function in a stably transfected cell line even if the gene of interest is essential. Also, induction of miR RNAi expression can be halted so phenotypic changes can be measured during recovery of gene function.

For a variety of expression options, the miR RNAi cassette, which contains EmGFP (pcDNA™6.2-GW/EmGFP-miR vector only), miR flanking regions, and an miRNA homologous to the target of interest, can be readily moved into a variety of DEST vectors. This occurs through Gateway™ recombination reactions in which the miR RNAi cassette is transferred into a pDONR™ vector (BP reaction) and then into a DEST vector (LR reaction) of choice. The HiPerform™ system has the new pLenti6.4/R4R2/V5-DEST vector for high virus titers and EmGFP expression.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodGateway
Constitutive or Inducible SystemConstitutive
Delivery TypeLentiviral
No. of Reactions20 Reactions
Product LineBLOCK-iT, HiPerform
Product TypePol II miR RNAi Expression Vector Kit
PromoterEF-1α, CMV
Protein TagV5 Epitope Tag
Quantity20 reactions
RNAi TypeExpressed RNAi (miRNA, shRNA)
Reporter GeneGFP (EmGFP)
Selection Agent (Eukaryotic)Blasticidin
Selection Marker PromoterPGK Promoter
VectorpLenti
Unit SizeEach
Contents & Storage
The BLOCK-iT™ HiPerform™ Lentiviral PolII miR RNAi Expression System with EmGFP contains the BLOCK-iT™ Pol II miR RNAi Expression Vector Kit with EmGFP plus all the necessary reagents for Gateway™ cloning into the lentiviral expression system, including: pLenti6.4/R4R2/V5-DEST vector, pLenti6.4/CMV/V5-MSGW/lacZ Control vector, pDONR™221, pENTR5’/EF1αp, pENTR5’/CMVp, ViraPower™ Lentiviral Bsd Support Kit (ViraPower™ Packaging Mix, Lipofectamine™ 2000 transfection reagent, and Blasticidin), 293FT Cell Line, One Shot™ Stbl3™ Chemically Competent E. coli, One Shot™ TOP10 Chemically Competent E. coli, and Gateway™ LR and BP Clonase™ II Enzyme Mixes. Store the Lipofectamine™ 2000 reagent at +4°C (do not freeze). Store the vectors, ViraPower™ Packaging Mix, Blasticidin, and both Clonase™ II Enzyme Mixes at -20°C. Store the competent cells at -80°C. Store the 293FT Cell Line in liquid nitrogen. All reagents are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

I'm getting no fluorescence signal with my expression clone containing EmGFP. What should I do?

Please ensure that the recommended filter sets for detection of fluorescence are used. Use an inverted fluorescence microscope for analysis. If desired, allow the protein expression to continue for 1-3 days before assaying for fluorescence.

I'm seeing nonspecific, off-target gene knockdown. What should I do?

The target sequence used may contain strong homology to other genes; please select a different target region.

I am not getting any colonies after titering. What would suggest I try?

Perform a kill curve to determine the antibiotic sensitivity of your cell line. Ensure that viral stocks are stored properly at -80 degrees C, and do not undergo freeze/thaw more than 3 times. Lastly, transducer the lentiviral contruct into cells in the presence of Polybrene reagent.

I'm getting few or no colonies, even with the transformation control. What could be the cause of this?

Ensure that the competent cells used were stored properly at -80 degrees C, and thawed on ice for immediate use. When adding DNA, mix competent cells gently: do not mix by pipetting up and down. Also do not exceed the maximum recommended amount of DNA for transformation (100 ng) or allow the volume of DNA added to exceed 10% of the volume of the competent cells, as these may inhibit the transformation.

I'm getting differently sized colonies after TOP10 E. coli transformation when using the miRNA lentiviral expression system. Which one should I use?

Some transformants may contain plasmids in which unwanted recombination has occurred between the 5' and 3' LTR. We recommend using our One Shot Stbl3 Chemically Competent E. coli cells, as they help in stabilizing lentiviral DNA containing direct repeats, and generally give rise to fewer unwanted recombinants. We recommend screening both colony sizes; however, in general, for lentiviral plasmids the small colonies tend to be the correct clones. Large colonies may have undergone a recombination event to delete part of the plasmid, thus allowing the cells to grow faster.