Product Image
Invitrogen™
BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP
The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits combine the advantages of traditional RNAi vectors (stable expression and the ability to use viral delivery) with capabilities for tissue-specific expression and multiple target knockdown from the same transcript.
Have Questions?
Catalog NumberQuantity
K493900
also known as K4939-00
20 Reactions
Catalog number K493900
also known as K4939-00
Price (USD)
2,280.00
Each
Quantity:
20 Reactions
Price (USD)
2,280.00
Each
The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits combine the advantages of traditional RNAi vectors (stable expression and the ability to use viral delivery) with capabilities for tissue-specific expression and multiple target knockdown from the same transcript. The BLOCK-iT™ Pol II miR RNAi Expression Vector Kits and the BLOCK-iT™ Lentiviral Pol II miR RNAi Expression Systems are designed to express artificial miRNAs which are engineered to have 100% homology to your target sequence and will result in target cleavage. Using Invitrogen's award-winning BLOCK-iT™ RNAi Designer, over 70% of constructs produce more than 70% knockdown.
The Invitrogen Pol II expression family of vectors include:
  • Strong expression from the CMV immediate early promoter, with the option to use tissue-specific or other regulated promoters via MultiSite Gateway™ recombination.
  • Compatibility with many of Invitrogen's Gateway™ destination (DEST) vectors for gene expression.
  • Ability to control the initiation of the RNAi response with a new BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit w/EmGFP.
  • A new destination vector in the BLOCK-iT™ HiPerform™ Lentiviral PolII miR RNAi Expression System with EmGFP. The new vector contains an mRNA stabilizing sequence (WPRE) and a nuclear import sequence (cPPT) which generate up to 5-fold higher virus titers and EmGFP expression levels. Additionally, blasticidin resistance is expressed from the mouse Pgk-1 promoter to avoid shut-down after multiple passages.
  • Co-cistronic expression of Emerald GFP (EmGFP), resulting in correlation of EmGFP expression with knockdown from your miR RNAi.
  • Expression of more than one engineered miR RNAi sequence on the same transcript, allowing the knockdown of multiple genes simultaneously and the generation of synthetic phenotypes
The new BLOCK-iT™ Inducible Pol II miR RNAi Expression Vector Kit with EmGFP provides the ability to regulate RNAi experiments. This kit contains the pT-REx-DEST30 Gateway™ vector which after simple cloning and shuttling techniques, produces a miR RNAi expression vector suitable for inducible knockdown. The pT-REx-DEST30 Gateway™ vector contains the CMV promoter with two copies of the tetracycline operator (tetO2) sequence allowing high-level and regulated expression. This permits the study of loss of function in a stably transfected cell line even if the gene of interest is essential. Also, induction of miR RNAi expression can be halted so phenotypic changes can be measured during recovery of gene function.
For a variety of expression options, the miR RNAi cassette, which contains EmGFP (pcDNA™6.2-GW/EmGFP-miR vector only), miR flanking regions, and an miRNA homologous to the target of interest, can be readily moved into a variety of DEST vectors. This occurs through Gateway™ recombination reactions in which the miR RNAi cassette is transferred into a pDONR™ vector (BP reaction) and then into a DEST vector (LR reaction) of choice. The HiPerform™ system has the new pLenti6.4/RF R2/V5-DEST destination vector for high virus titers and EmGFP expression.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodGateway™
Constitutive or Inducible SystemInducible
Delivery TypeTransfection
GeneGFP (EmGFP)
Product LineBLOCK-iT
Product TypeRNAi Expression Vector Kit
Quantity20 Reactions
RNAi TypemiRNA
Selection Agent (Eukaryotic)Blasticidin
VectorBLOCK-iT RNAi Vectors
FormatKit
PromoterCMV/TO
Unit SizeEach
Contents & Storage
BLOCK-iT™ Inducible PolII miR RNAi Expression Vector Kit w/EmGFP
A cloning box with:
• Linearized pcDNA™6.2-GW/EmGFP-miR
• Annealing buffer (10X)
• T4 DNA Ligase
• DNA Ligation buffer (5X)
• lacZ control oligo
• lacZ control plasmid
• Negative control plasmid
• DNase/RNase-free water
• Forward and reverse sequencing primers
pT-REx-DEST30 Gateway™ Vector Kit
• pT-REx-DEST30
• pT-REx/GW-30/lacZ vectors
T-REx™ Regulatory Module
• pcDNA™6/TR
• Tetracycline
The Gateway™ LR and BP Clonase™ kits, blasticidin, and pDONR™ 221 are also included.
Store the vectors, buffers, control oligo and plasmids, antibiotics, clonase kits, water, and sequencing primers at -20°C. The One Shot™ box contains transformation reagents including twenty-one 50-μL aliquots of One Shot™ TOP10 Chemically Competent E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid. Store these transformation reagents -80°C.
Frequently asked questions (FAQs)
What are the benefits of the BLOCK-iT miR RNAi expression system over similar systems?
The BLOCK-iT miR RNAi expression system allows you to take advantage of promoter flexibility by choosing from a variety of Pol II promoters like CMV, Ubc, tissue specific, or inducible promoters. The miRNA vectors also allow you to clone multiple sequences in the same vector, thereby enabling you to target more than one gene or more than one location in a gene using a single plasmid. An additional advantage offered by some of the miRNA expression vectors is that transfection efficiency can be monitored with the EmGFP fusion partner.
Find additional tips, troubleshooting help, and resources within our RNAi Support Center.
How can I tell which sequence will be a potent miRNA sequence?
We offer a free online RNAi design program to help you design an effective miRNA sequence. The RNAi designer can be found at https://rnaidesigner.invitrogen.com/rnaiexpress/, or search "RNAi Designer" from the homepage of thermofisher.com.
Can I target multiple genes or multiple locations within a gene using the BLOCK-iT miR RNAi expression system?
Yes. The miRNA cloning vectors are designed such that you can easily chain together multiple miRNAs to express them in one transcript. This can be used to increase transcription levels of the same miRNA sequence, or to combine multiple different sequences in one co-cistronic expression cassette.
Are the BLOCK-iT miR RNAi Expression Kits compatible with adenoviral expression systems?
Yes. The miR miRNA vectors are Gateway cloning compatible, and you could use Gateway cloning to transfer the miR miRNA expression cassette to any of our Gateway-adapted viral expression vectors.
How do the BLOCK-iT miR RNAi Kits work? Do they cleave the specific mRNA target or just suppress translation? Can this system be used for analyzing endogenous miRNAs?
The miRNA vectors use endogenous miRNA processing machinery to allow targeted RNAi knockdown of specific genes, but they are not tested for use in the analysis of endogenous miRNA. Endogenous miRNAs are typically not 100% homologous to their targets and thus invoke translational suppression. Our products are designed with 100% homology to the target gene and generally result in target mRNA cleavage.
Citations & References (1)
Citations & References
Abstract
AID-induced decrease in topoisomerase 1 induces DNA structural alteration and DNA cleavage for class switch recombination.
Authors:Kobayashi M, Aida M, Nagaoka H, Begum NA, Kitawaki Y, Nakata M, Stanlie A, Doi T, Kato L, Okazaki IM, Shinkura R, Muramatsu M, Kinoshita K, Honjo T,
Journal:Proc Natl Acad Sci U S A
PubMed ID:20018730
'To initiate class switch recombination (CSR) activation-induced cytidine deaminase (AID) induces staggered nick cleavage in the S region, which lies 5'' to each Ig constant region gene and is rich in palindromic sequences. Topoisomerase 1 (Top1) controls the supercoiling of DNA by nicking, rotating, and religating one strand of DNA. ... More