Product Image
Invitrogen™

ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit

The ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit.
Have Questions?
Catalog NumberQuantity
K533000
also known as K5330-00
1 kit
Catalog number K533000
also known as K5330-00
Price (USD)
3,455.00
Each
Quantity:
1 kit
Price (USD)
3,455.00
Each

The ViraPower™ HiPerform™ Lentiviral Gateway™ Expression Kit includes all the components needed to generate lentivirus, including vector kit, 293FT cell line, and the support kit. This kit combines Invitrogen's™ ViraPower™ HiPerform™ Lentiviral and Gateway™ technologies to facilitate easy recombination-based cloning and lentiviral-based, high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti6.3/V5-DEST™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements.

Advantages

  • Stable expression
  • Long-term experiments
  • Accurate titer of functional virus
  • Flexible and versatile Gateway™ recombination cloning technology

Key Features

  • HiPerform™ WPRE and cPPT elements
  • CMV promoter
  • V5 epitope tag at C terminus
  • Blasticidin selection

Kit includes

  • pLenti6.3/V5-DEST™ Gateway™ Vector Kit (V533-06)
  • ViraPower™ Bsd Lentiviral Support Kit (K4970-00)
  • 293FT Cell Line (R700-07)

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Cloning MethodGateway
Constitutive or Inducible SystemConstitutive
Delivery TypeLentiviral
For Use With (Application)Protein Expression
For Use With (Equipment)HiPerform™, TOPO ™ , ViraPower™
Key FunctionsViral Expression
Product LineGateway, HiPerform, ViraPower
Product TypeExpression Kit
Protein TagV5 Epitope Tag
Quantity1 kit
Selection Agent (Eukaryotic)Blasticidin
Special ElementsWPRE, cPPT
Cell Type293-FT
FormatKit
PromoterCMV
VectorpLenti, TOPO-TA Vector
Unit SizeEach
Contents & Storage

Vector kit
• pLenti6.3/V5-DEST™ (150 ng/μL), 40 μL, store at -20°C
• pLenti6.3/V5-GW/lacZ Control vector (0.5 μg/μL), 20 μL, store at -20°C
• One Shot™ Stbl3™ Chemically Competent E. coli, 21 x 50 μL, store at -80°C
• pUC19 control DNA (10 pg/μL), 50 μL, store at -80°C
• S.O.C. Medium, 6 mL, store at -80°C

Support kit
• ViraPower™ Packaging Mix, 195 μg, store at -20°C
• Lipofectamine™ 2000, 0.75 mL, store at 4°C
• Blasticidin, 50 mg, store at -20°C
• 293 FT cells (3 x 106 cells/mL), 1 mL, store in liquid nitrogen

Frequently asked questions (FAQs)

Can I perform the single-step protocol for the BP/LR Clonase reaction using BP Clonase enzyme and LR Clonase enzyme instead of BP Clonase II enzyme and LR Clonase II enzyme?

In the single-step protocol for the BP/LR Clonase reaction, we would not recommend substituting the BP Clonase II/LR Clonase II enzymes with BP Clonase /LR Clonase enzymes as this would result in very low recombination efficiency.

Do you have a recommended single-step protocol for BP/LR recombination?

Yes, we have come up with a single-step protocol for BP/LR Clonase reaction (http://www.thermofisher.com/us/en/home/life-science/cloning/gateway-cloning.html#1), where DNA fragments can be cloned into Destination vectors in a single step reaction, allowing you to save time and money.

How can I move my gene of interest from a Gateway-adapted expression clone to a new Destination vector as I have lost the entry clone?

We would recommend performing a BP reaction with a Donor vector in order to obtain an entry clone. This entry clone can then be used in an LR reaction with the Destination vector to obtain the new expression clone.

Can I purchase the 5X LR Clonase buffer or 5X BP Clonase buffer separately?

We do not offer the 5X LR Clonase buffer and 5X BP Clonase buffer as standalone products. They are available as part of the enzyme kits.

Do you offer Gateway vectors for expression in plants?

We do not offer any Gateway vectors for expression in plants.