Thermo Scientific™
IDEIA™ Borrelia burgdorferi IgM Kit using Enzyme Immunoassay
Catalog number: K603011-2
Have Questions?
Thermo Scientific™
IDEIA™ Borrelia burgdorferi IgM Kit using Enzyme Immunoassay
Catalog number: K603011-2
Have Questions?
Rapidly detect the human serum IgM antibodies to Borrelia burgdorferi with Thermo Scientific™ IDEIA™ Borrelia burgdirferi IgM Kit. The enzyme immunoassay is designed to deliver rapid detection of human serum IgM antibodies to Borrelia burgdorferi sensu lato as an aid in the diagnosis of Lyme borreliosis.
Catalog Number
K603011-2
Unit Size
Each
Price (USD)
Full specifications
DescriptionIDEIA™ Borrelia burgdorferi IgM Enzyme Immunoassay
Detectable AnalytesIgM antibodies to Borrelia burgdorferi
TypeBorrelia Testing Kit
IncludesMicrowells, sample diluent, wash buffer concentrate, IgM cut-off control, IgM positive control, flagellum conjugate, peroxidase and reconstitution buffer, substrate, stop solution, instruction booklet
Quantity46 Tests
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
K603011-2Full specifications
EachRequest A Quote
DescriptionIDEIA™ Borrelia burgdorferi IgM Enzyme Immunoassay
Detectable AnalytesIgM antibodies to Borrelia burgdorferi
TypeBorrelia Testing Kit
IncludesMicrowells, sample diluent, wash buffer concentrate, IgM cut-off control, IgM positive control, flagellum conjugate, peroxidase and reconstitution buffer, substrate, stop solution, instruction booklet
Quantity46 Tests
Unit SizeEach
Showing 1 of 1
Lyme borreliosis is a multisystem infection caused by the tickborne spirochete Borrelia burgdorferi sensu lato1,2. Due to the extreme paucity of B. burgdorferi in pathological lesions and body fluids, culture and techniques for direct detection of spirochetal DNA may not be successful in the routine diagnosis of Lyme borreliosis. The measurement of B. burgdorferi–specific antibodies is a suitable method.
Use IDEIA Borrelia burgdorferi IgM Kit for detection of human serum IgM antibodies to Borrelia burgdorferi sensu lato.
  • Simple—ready-to-use liquid reagents and concentrates make the assay easy to implement and run
  • Excellent sensitivity and specificity—high quality, well characterized antibodies used in the assay provide excellent sensitivity and specificity
  • Easy-to-read results
IDEIA Borrelia burgdorferi IgM Kit uses purified, native B. afzelii strain DK1 flagellum as test antigen. The flagellum is highly immunogenic and elicits an early, strong and persistent immune response3,4. Compared with the conventional and currently most widely used test antigen, which is based on whole cell extract of the spirochete, the purified, native flagellar antigen improves the diagnostic sensitivity and specificity of serological assays for Lyme borreliosis5,6. Furthermore, the use of flagellum as test antigen is appropriate in all geographical areas as the flagellum shows no significant variation between B. burgdorferi strains.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  