IDEIA™ PCE Chlamydia Kit using Enzyme Immunoassay
Thermo Scientific™
IDEIA™ PCE Chlamydia Kit using Enzyme Immunoassay
Carry out rapid detection of Chlamydia antigen in clinical samples with this dual amplification assay.
Have Questions?
Número do catálogo K603211-2
Preço (BRL)
-
Carry out rapid detection of Chlamydia antigen in clinical samples with Thermo Scientific™ IDEIA™ PCE Chlamydia Kit – an immunoassay utilizing dual amplification technology for detection of Chlamydia antigen in human urethral and endocervical swabs and male urine samples. The test utilizes Chlamydia genus-specific monoclonal antibody of established specificity. The presence of Chlamydia antigen in clinical specimens is indicated by the colored end point.
Chlamydia trachomotis is one of the most prevalent pathogens in the world causing a variety of human infections. It is also the most common cause of sexually transmitted diseases 1, 2.
Use IDEIA PCE Chlamydia Kit as a sensitive, specific and economical means of routinely diagnosing Chlamydial infections.3, 4, 5
  • Rapid- results within 3 minutes
  • Simple- ready-to-use liquid reagents and concentrates make the assay easy to implement and run
  • Excellent sensitivity and specificity- high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity.
  • Easy-to- read results
Diagnostic methods for detection of Chlamydia include traditional culture techniques which are dependent upon the presence of viable elementary bodies in clinical specimens. Elementary bodies undergo replication in cell culture and the inclusions formed are detected microscopically using conventional or immunofluorescence staining techniques. Most commonly, diagnosis of Chlamydia infection is performed either by direct detection of Chlamydia elementary bodies in clinical specimens using immunofluorescent monoclonal antibody reagents (e.g. IMAGEN™ Chlamydia) or by detection of Chlamydia antigen by immunoassays3. Other methods include techniques such as the polymerase chain reaction for detection of Chlamydia nucleic acids4. Culture techniques are expensive and require specialized laboratory facilities. Methods for detection of nucleic acids also require specialized equipment and laboratory facilities to minimize specificity problems. Direct detection by immunofluorescence staining becomes labor intensive when used to routinely screen large numbers of specimens. Amplified immunoassay techniques for antigen detection (e.g. IDEIA PCE Chlamydia) provide a sensitive, specific and economical means of routinely diagnosing Chlamydial infections.3, 4, 5
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
1.Chlamydia Disease (1983) Ed. Darougar, S. British Medical Bulletin 39: 107-203
2. Schachter, J. und Dawson, C. R. (1979) Psittacosis-lymphogranuloma venereum agents/TRIC agents. In: Lennette, E. H. und Schmidt, N. J. Hrsg. Diagnostic procedures for viral, rickettsial and chlamydial infections. 5. Auflage, American Public Health Association, 1021-1059
3. Hirose T, Iwasawa Am Satoh T, Itoh N, Tsukamoto T, Gohro T, Miyagusgu T, Ikegaki S, Saka T, Nishimura M, Yamazaki K, Yoshida H, Hagiwara T Clinical study of the effectiveness of a dual amplified immunoassay (IDEIA PCE Chlamydia) for the diagnosis of male urethritis. Int J STD AIDS 1998 Jul; 9 (7): 414-417
4. Takashi Deguchi, Mitsuru Yasduda, Masahiro Uno, Kohji Tada, Hideki Iwata, Hisao Komeda, Shin-Ichi Maeda, Vivian Latila, Isao Saito, Yukimichi Kawada. Comparison among performance of a ligase chain reaction-based assay and two enzyme immunoassays in detecting Chlamydia trachomatis in urine specimens from men with Nongonococcal Urethritis. Journal of Clinical Microbiology, July 1996, 1708 – 1710
5. Paul I, Leece J, Caul E O. Chlamydia trachomatis: a review of its laboratory diagnosis and apreliminary evaluation of a new DNA-based assay. PHLS Microbiology Digest 13 (4)
Especificações
DescriptionIDEIA PCE Chlamydia
Detectable AnalytesChlamydia antigen
TypeChlamydia Testing Kit
IncludesTwo 96 well microtitration plates, transport medium (x10), positive control, negative control, 2x polymer conjugate, wash buffer concentrate (x20), 2x amplifier A, 2x amplifier B, 2x stop solution, instruction booklet
Quantity192 Tests
Unit SizeEach