IMAGEN™ Herpes Simplex Virus (HSV) Kit using  Direct Immunofluorescence Assay
Thermo Scientific™
IMAGEN™ Herpes Simplex Virus (HSV) Kit using Direct Immunofluorescence Assay
Carry out rapid detection and identification of HSV1 and HSV2 in cell cultures with this direct immunofluorescence assay.
Have Questions?
Catalog NumberQuantity
K610611-250 Tests
Catalog number K610611-2
Price (USD)
-
Quantity:
50 Tests
The IMAGEN™ Herpes Simplex Virus typing test is a qualitative direct immunofluorescence test for the detection and typing of HSV 1 and HSV 2 in cell cultures.
HSV is a common and universal infection of humans, associated with a wide range of clinical diseases in both immunocompetent and immunocompromised individuals. Both HSV 1 and 2 are frequently implicated in localised vesicular infections of the skin, conjunctiva and mucous membranes of the mouth or genitalia1, 2,3.
Use IMAGEN Herpes Simplex Virus (HSV) Kit as a rapid, sensitive and specific method for the detection and differentiation of HSV1 and HSV2 isolates in cell culture monolayers.
  • Rapid results- within little over 30 minutes of receiving test sample or culture
  • Simple- ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity- high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity.
  • Easy-to-read results
  • Long Shelf Life- typically 24 months.
Herpes simplex virus is a DNA virus containing an icosahedral nucleocapsid surrounded by a lipid containing envelope. Human HSV is classified within the family Herpesviridae and is a member of the sub-family Alphaherpesvirinae. The genus Simplexvirus includes two type species of human herpes simplex virus, HSV 1 and HSV 24.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
General References:
  1. Herpes Simplex Virus infections. In Herpes virus infections: clinical aspects (ed. Glaser, R.) Marcel Dekker Inc., New York, pp 1-55.
  2. Longson M. (1990) Herpes simplex. In principles and practice of clinical virology (eds. A.J.Zuckerman et al) John Wiley and Sons Ltd, pp 3-42.
  3. Corey L. and Spear P.G. (1986) Infections with Herpes Simplex Virus Part 2. New England Journal of Medicine 314: No 12: 749 757.
  4. Francki R.I.B., Fauquet C.M., Knudson D.L. and Brown F. Classification and nomenclature of viruses. Fifth Report of the International Committee on Taxonomy of Viruses. Archives of Virology, Supplement 2, Spurger Velacy, New York, pp 103-106.
Specifications
DescriptionIMAGEN Herpes Simplex Virus (HSV) Kit
Includes2 x 2 positive control slide, mounting fluid, HSV type 1 reagent, HSV type 2 reagent, instruction for use
Quantity50 Tests
Detectable AnalytesHSV 1/2
Unit SizeEach
Contents & Storage
2°C to 8°C