Product Image
Invitrogen™

InsectSelect™ Glow Kit with Sf9 Cells

The InsectSelect™ Glow Kit features the pIZT/V5-His vector for high-level expression of yourgene of interest. This vector has the following深入閱讀
Have Questions?
產品號碼Quantity
K81001
亦稱為 K810-01
1 kit
產品號碼 K81001
亦稱為 K810-01
價格 (TWD)
84,550.00
線上優惠
Ends: 30-Sep-2026
105,687.00
您節省 21,137.00 (20%)
Each
Quantity:
1 kit
價格 (TWD)
84,550.00
線上優惠
Ends: 30-Sep-2026
105,687.00
您節省 21,137.00 (20%)
Each
The InsectSelect™ Glow Kit features the pIZT/V5-His vector for high-level expression of your
gene of interest. This vector has the following features:

• The OpIE2 promoter for constitutive expression
• The Zeocin™ resistance gene for rapid selection of stably transfected cell lines
• C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Invitrogen's Anti-V5 Antibody and rapid purification with nickel-chelating resin

In addition, pIZT/V5-His expresses a fusion of the green fluorescent protein and the Zeocin™ resistance
protein (Zeo-GFP). The Zeo-GFP fusion protein permits rapid selection of stably transfected cell lines
with Zeocin™ and confers a fluorescent phenotype that simplifies identification of transfected cells.
For Research Use Only. Not for use in diagnostic procedures.
規格
Product TypeSf9 Cells Glow Kit
Protein Tag Position (to your gene)C-terminal
Quantity1 kit
VectorpIZ
Cloning MethodRestriction Enzyme/MCS
Product LineInsectSelect
PromoterOplE2
Protein TagHis Tag (6x), V5 Epitope Tag
Unit SizeEach
內容物與存放
The InsectSelect™ Glow Kit includes 20 μg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2 μg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1 g Zeocin,™ Cellfectin™ Reagent, Sf9 cells, and 1 L of GIBCO™ Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described above. Store frozen insect cells in liquid nitrogen. Store medium and Cellfectin™ Reagent at +4°C. Store all other reagents at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

常見問答集 (常見問題)

Does the incubator need to be humidified when culturing insect cells?

Humidification is only necessary for monolayer cultures and suspension cultures with volumes less than 35 mL. With shaker or spinner cultures greater than 35 mL, humidification is not necessary.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

引用資料與參考文獻 (2)

引用資料與參考文獻
Abstract
Disarming the mustard oil bomb.
Authors: Ratzka Andreas; Vogel Heiko; Kliebenstein Daniel J; Mitchell-Olds Thomas; Kroymann Juergen;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12161563
Plants are attacked by a broad array of herbivores and pathogens. In response, plants deploy an arsenal of defensive traits. In Brassicaceae, the glucosinolate-myrosinase complex is a sophisticated two-component system to ward off opponents. However, this so-called  ... More
Punctin, a novel ADAMTS-like molecule, ADAMTSL-1, in extracellular matrix.
Authors: Hirohata Satoshi; Wang Lauren W; Miyagi Masaru; Yan Lin; Seldin Michael F; Keene Douglas R; Crabb John W; Apte Suneel S;
Journal:J Biol Chem
PubMed ID:11805097
Punctin (ADAMTSL-1) is a secreted molecule resembling members of the ADAMTS family of proteases. Punctin lacks the pro-metalloprotease and the disintegrin-like domain typical of this family but contains other ADAMTS domains in precise order including four thrombospondin type I repeats. Punctin is the product of a distinct gene on human ... More