Product Image
Invitrogen™
Bac-N-Blue™ Transfection Kit
The Bac-N-Blue™ Linear DNA was specifically designed for recombination with the pBlueBac vectors and pMelBac. Recombinant viruses have a full-length,Read more
Have Questions?
Catalog NumberQuantity
K85501
also known as K855-01
5 transfections
Catalog number K85501
also known as K855-01
Price (USD)
1,146.00
Each
Quantity:
5 transfections
Price (USD)
1,146.00
Each
The Bac-N-Blue™ Linear DNA was specifically designed for recombination with the pBlueBac vectors and pMelBac. Recombinant viruses have a full-length, functional lacZ gene resulting in the production of
blue plaques. This allows for easy identification and purification. Bac-N-Blue™ DNA can be used with any polyhedrin promoter-based baculovirus transfer vector. Bac-N-Blue™ DNA is linearized at three
sites, one of which is in a gene that is essential for viral propagation (1). This leads to a decrease in non-recombinant virus, making selection and purification of recombinant virus easy. In addition to the viral DNA, the Bac-N-Blue™ Transfection Kit contains Cellfectin™ Reagent for high transfection efficiency.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeInsect Cell Transfection Kit
Quantity5 transfections
VectorpBlueBac
Cloning MethodRestriction Enzyme/MCS
Product LineBac-N-Blue
PromoterPolyhedrin
Protein TagUntagged
Unit SizeEach
Contents & Storage
The Bac-N-Blue™ Transfection Kit contains five 0.5 μg vials of Bac-NBlue™ linear virus DNA and 125 μl of Cellfectin™ Reagent.
Store at +4°C. All components are guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
I cannot grow this white colony in liquid culture. What should I do?
The concentration of gentamicin might be too high. Try lowering the amount to 5 µg/mL and try adding more of the colony to the culture medium.
What has happened when I see blue colonies? How about colonies which are blue in the center and white on the edges?
In the case of a blue colony, the E. coli has the bacmid and the plasmid in it, allowing the cells to survive the selection process. However, because the transposition has not occurred, the LacZ gene is not disrupted. For bulls-eye colonies, this indicates that the transposition took place when the colony was growing. Re-streaking for an isolated clone from the white portion of the mixed colony should yield some colonies where transposition occurred.
I'm getting mostly white/wild-type plaques instead of blue/recombinant plaques. What am I doing wrong?
This is typically an indication of poor homologous recombination. Check the plasmid/linear DNA ratio you used. If there are some blue plaques, however, expand those viruses and check for their protein. In our experience, they are correct, even if they were in relatively low abundance.
I've infected my cells and see large polyhedra in one cell and smaller polyhedra (more numerous) in a neighboring cell. Is this normal?
Yes, cells are infected with wild-type virus individually and will develop polyhedra at different rates until all the cells in the flask are infected. The polyhedra in cells will form in approximately 3-4 days, differing in size and number until they reach their maximum capacity and burst the cell, releasing tiny particles of virus into the medium.
I'm worried that I am not getting plaques. How many days does it take to see plaques and what size are they typically?
Normally, very small white dots show up about 5-7 days and 1 mm plaques show up around day 10. Plaques can vary in size from 1 mm to 4 mm.
Citations & References (26)
Citations & References
Abstract
OX40 stimulation by gp34/OX40 ligand enhances productive human immunodeficiency virus type 1 infection.
Authors: Takahashi Y; Tanaka Y; Yamashita A; Koyanagi Y; Nakamura M; Yamamoto N;
Journal:J Virol
PubMed ID:11435553
'OX40 is a member of the tumor necrosis factor (TNF) receptor superfamily and known to be an important costimulatory molecule expressed on activated T cells. To investigate the role of costimulation of OX40 in human immunodeficiency virus type 1 (HIV-1) infection by its natural ligand, gp34, the OX40-transfected ACH-2 cell ... More
Identification of the major synaptojanin-binding proteins in brain.
Authors:de Heuvel E, Bell AW, Ramjaun AR, Wong K, Sossin WS, McPherson PS
Journal:J Biol Chem
PubMed ID:9079704
'Synaptojanin is a nerve-terminal enriched inositol 5-phosphatase thought to function in synaptic vesicle endocytosis, in part through interactions with the Src homology 3 domain of amphiphysin. We have used synaptojanin purified from Sf9 cells after baculovirus mediated expression in overlay assays to identify two major synaptojanin-binding proteins in rat brain. ... More
Identification of a novel human eicosanoid receptor coupled to G(i/o).
Authors: Hosoi Takeshi; Koguchi Yutaka; Sugikawa Emiko; Chikada Aiko; Ogawa Koji; Tsuda Naoki; Suto Naoki; Tsunoda Shiho; Taniguchi Tomoyasu; Ohnuki Tetsuo;
Journal:J Biol Chem
PubMed ID:12065583
'We have conducted an in silico data base search for and cloned a novel G-protein-coupled receptor (GPCR) named TG1019. Dot and Northern blotting analyses showed that transcripts of the novel GPCR were expressed in various tissues except brain, and the expression was more intense in liver, kidney, peripheral leukocyte, lung, ... More
VRILLE feeds back to control circadian transcription of Clock in the Drosophila circadian oscillator.
Authors:Glossop NR, Houl JH, Zheng H, Ng FS, Dudek SM, Hardin PE,
Journal:Neuron
PubMed ID:12546820
'The Drosophila circadian oscillator consists of interlocked period (per)/timeless (tim) and Clock (Clk) transcriptional/translational feedback loops. Within these feedback loops, CLK and CYCLE (CYC) activate per and tim transcription at the same time as they repress Clk transcription, thus controlling the opposite cycling phases of these transcripts. CLK-CYC directly bind ... More
Accurate processing and secretion in the baculovirus expression system of an erythroid-cell-stimulating factor consisting of a chimaera of insulin-like growth factor II and an insect insulin-like peptide.
Authors:Congote LF, Li Q
Journal:Biochem J
PubMed ID:8166627
'A synthetic gene encoding the signal peptide and the N-terminal sequence of bombyxin, an insect insulin-like peptide, and the 58 amino acids of the C-terminal sequence of human insulin-like growth factor II (IGF II) has been expressed using the baculovirus system. This synthetic chimaera was obtained by amplification of four ... More