본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
Product Image
Invitrogen™
FreeStyle™ 293 Expression System
The FreeStyle™ 293 Expression System is a complete, suspension cell culture system for generating large amounts of mammalian recombinant protein.자세히 알아보기
Have Questions?
카탈로그 번호수량
K9000011 Kit
카탈로그 번호 K900001
제품 가격(KRW)
1,757,000
온라인 행사
Ends: 30-Jun-2026
2,066,000
할인액 309,000 (15%)
Each
수량:
1 Kit
제품 가격(KRW)
1,757,000
온라인 행사
Ends: 30-Jun-2026
2,066,000
할인액 309,000 (15%)
Each
The FreeStyle™ 293 Expression System is a complete, suspension cell culture system for generating large amounts of mammalian recombinant protein. Free your lab from the time-consuming, labor-intensive process of seeding, feeding, passing, and maintaining multiple flasks of adherent cells (Figure 1). The FreeStyle™ 293 Expression System combines GIBCO™ FreeStyle™ 293 Expression Medium with 293fectin™, a cationic-lipid formulation designed to transfect suspension FreeStyle™ 293-F cells at high efficiency. FreeStyle™ 293-F cells and 293fectin™ are available as part of the Expression System or separately.
FreeStyle™ 293 Expression Medium is chemically-defined, protein-free medium specifically developed for the ability to support the growth and transfection of 293-F cells under suspension type culture conditions (Figure 2). The medium is a complete, ready-to-use medium that has been supplemented with GlutaMAX™-I Supplement and is animal-origin free. FreeStyle™ 293 Expression Medium is able to save significant time and costs associated with adaptation of cell cultures to serum-free conditions.
For Research Use Only. Not for use in diagnostic procedures.
사양
세포주FreeStyle™ 293-F Cells
분류Serum-Free
농도1 x 10^7 cells/mL
배양 유형Suspension Cell Culture
배달 유형Transfection
발현 메커니즘Cell-Based Expression
발현 시스템Mammalian
용도(애플리케이션)Protein Expression
주요 기능Protein Production
제품라인Freestyle
제품 유형Expression System
수량1 Kit
셀 유형293-derived Cells
Unit SizeEach
구성 및 보관
The FreeStyle™ 293 Expression System contains:
  • 1L GIBCO™ FreeStyle™ 293 Expression Medium
  • 1 x 107 FreeStyle™ 293-F Cells (frozen)
  • 1 mL 293fectin™ Transfection Reagent
  • 100 mL Opti-MEM™ I for Complex Formation
  • 25 μg pCMV-SPORT β-gal
Please review the FreeStyle™ 293 System Manual for storage and handling information. Store FreeStyle™ 293-F Cells in liquid nitrogen. Store 293fectin™ Transfection Reagents at 4°C. Store GIBCO™ FreeStyle™ 293 Expression Medium and Opti-MEM™ I at 2-8°C in the dark. Store pCMV SPORT β-gal at -20°C. Reagents are guaranteed stable for six months when stored properly.
자주 묻는 질문(FAQ)
I am using your FreeStyle Max 293 Expression System and my cell viability is at most 87-90%. Is this normal? Do you have any tips to increase the cell viability? I am using 125 mL and 250 mL non-baffled, polycarbonate shaker flasks with vented caps. Will using baffled flasks help? Should I add Pluronic reagent to the medium?
Culture viability should be in the high 90% range for the seed stock that you are maintaining and growing for the transfections. After transfection, the viability will slowly decrease to the lower 90% to 80% range by about 3-6 days post-transfection and can drop more dramatically at 7 days or longer. This is when using our recommended protocol, which does not suggest any media changes or supplementation post-transfection.
A few parameters we recommend checking to assure high culture viability are: pH at ˜7.0, no visible clumping of the cells, CO2 levels at approximately 7-8% (this should be adjusted to achieve the appropriate pH), volume of culture at 40% or less of nominal flask capacity, and incubator temperature at 37 degrees C. If you have a shaker platform setup in your incubator, the platform may generate sufficient heat to increase the temperature of the incubator. We recommend using a thermometer attached to the shaker platform. Use of baffled flasks may help, if insufficient oxygenation of the cultures is suspected. We also recommend handling and storing the culture media properly, as light exposure can result in breakdown of certain media components. If you are using FreeStyle 293 Expression Medium, supplementing with Pluronic reagent is not necessary.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the main difference between the FreeStyle Max 293 Expression System and the FreeStyle 293 Expression System?
The two systems have the same components except for the transfection reagent and the medium supplied for formation of transfection complexes. The FreeStyle Max 293 Expression System contains FreeStyle Max Reagent and OptiPRO SFM (serum-free medium that is devoid of animal-origin components), whereas the FreeStyle 293 Expression System contains 293fectin Transfection Reagent and Opti-MEM I Reduced Serum Medium (serum-free medium but not animal-origin free).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Which vectors do you recommend for use with FreeStyle system?
Generally, any mammalian expression vector should be compatible with the FreeStyle expression system. Some of the more popular choices include pcDNA3.3-TOPO TA vector (Cat. No. K8300-01), pcDNA3.4-TOPO TA vector (Cat. no. A14697), pcDNA3.2 DEST vector (Cat. No. 12489018), pCEP vector (Cat. No. V044-50), or pcDNA4/HisMax vector (Cat. No. K864-20).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the difference between FreeStyle 293 Expression Medium and FreeStyle F17 Expression Medium?
FreeStyle F17 Expression Medium was designed from a different base formulation than FreeStyle 293 Expression Medium. FreeStyle F17 medium shows equal or better performance than the FreeStye 293 medium for certain cell lines and proteins. It is capable of supporting the growth of 293 and CHO cells and offers a wider variety and selection of transient cell culture media options.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
인용 및 참조 문헌 (5)
인용 및 참조 문헌
Abstract
Molecular dissection of the alpha-dystroglycan- and integrin-binding sites within the globular domain of human laminin-10.
Authors:Ido H, Harada K, Futaki S, Hayashi Y, Nishiuchi R, Natsuka Y, Li S, Wada Y, Combs AC, Ervasti JM, Sekiguchi K,
Journal:J Biol Chem
PubMed ID:14701821
'The adhesive interactions of cells with laminins are mediated by integrins and non-integrin-type receptors such as alpha-dystroglycan and syndecans. Laminins bind to these receptors at the C-terminal globular domain of their alpha chains, but the regions recognized by these receptors have not been mapped precisely. In this study, we sought ... More
Biological activity of follistatin isoforms and follistatin-like-3 is dependent on differential cell surface binding and specificity for activin, myostatin, and bone morphogenetic proteins.
Authors:Sidis Y, Mukherjee A, Keutmann H, Delbaere A, Sadatsuki M, Schneyer A,
Journal:Endocrinology
PubMed ID:16627583
Follistatin (FST) and FST-like-3 (FSTL3) are activin-binding and neutralization proteins that also bind myostatin. Three FST isoforms have been described that differ in tissue distribution and cell-surface binding activity, suggesting that the FST isoforms and FSTL3 may have some nonoverlapping biological actions. We produced recombinant FST isoforms and FSTL3 and ... More
Mutation of key residues of RPE65 abolishes its enzymatic role as isomerohydrolase in the visual cycle.
Authors:Redmond TM, Poliakov E, Yu S, Tsai JY, Lu Z, Gentleman S,
Journal:Proc Natl Acad Sci U S A
PubMed ID:16150724
RPE65 is essential for isomerization of vitamin A to the visual chromophore. Mutations in RPE65 cause early-onset blindness, and Rpe65-deficient mice lack 11-cis-retinal but overaccumulate alltrans-retinyl esters in the retinal pigment epithelium (RPE). RPE65 is proposed to be a substrate chaperone but may have an enzymatic role because it is ... More
Binding of anthrax toxin to its receptor is similar to alpha integrin-ligand interactions.
Authors:Bradley KA, Mogridge J, Jonah G, Rainey A, Batty S, Young JA,
Journal:J Biol Chem
PubMed ID:14507921
The secreted protein toxin produced by Bacillus anthracis contributes to virulence of this pathogen and can cause many of the symptoms seen during an anthrax infection, including shock and sudden death. The cell-binding component of anthrax toxin, protective antigen, mediates entry of the toxin into cells by first binding directly ... More
A novel secreted endonuclease from Culex quinquefasciatus salivary glands.
Authors:Calvo E, Ribeiro JM,
Journal:J Exp Biol
PubMed ID:16809456
Previous analysis of the salivary gland transcriptome of Culex quinquefasciatus showed the potential presence of an endonuclease with sequence similarities to shrimp, crab and two tsetse salivary proteins. Indeed, not only was the cloned cDNA shown to encode an active double-stranded endonuclease, but also the same activity was demonstrated to ... More