Search
| Catalog Number | Quantity |
|---|---|
| K95301 also known as K953-01 | 1 Kit |
The Ni-NTA Purification System is designed for the purification of recombinant proteins that contain a polyhistidine (6xHis) sequence. The kit utilizes Ni-NTA nickel-chelating resin and is supplied with native and denaturing buffers for efficient purification of recombinant proteins under different conditions.