µPAC™ Neo HPLC Columns
Thermo Scientific™

µPAC™ Neo HPLC Columns

Learn more about your proteome samples with Thermo Scientific™ μPAC™ Neo Plus HPLC columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesProduct Line
KIT-UPAC050NAN50 cm µPAC Neo Plus column, ACC-VSP1006, ACC-VSP2050, ACC-GRNDC, ES993µPAC Neo Plus
COL-NANO050NEOB50 cm μPAC™ Neo HPLC ColumnµPAC Neo
COL-NANO110NEOB110 cm μPAC™ Neo HPLC ColumnµPAC Neo
COL-CAPHTNEOB5.5 cm High Throughput μPAC™ Neo HPLC ColumnµPAC Neo
COL-UPAC005CAPHT µPAC Neo Plus columnµPAC Neo Plus
COL-UPAC050NAN50 cm µPAC Neo Plus columnµPAC Neo Plus
COL-UPAC110NAN110 cm µPAC Neo Plus columnµPAC Neo Plus
KIT-UPAC110NAN110 cm µPAC Neo Plus column, ACC-VSP1006, ACC-VSP2050, ACC-GRNDC, ES993µPAC Neo Plus
KIT-UPAC005CAPHT µPAC Neo Plus column, ACC-VSP2006, ACC-VSP2050, ACC-GRNDC, ES994µPAC Neo Plus
Catalog number KIT-UPAC050NAN
Price (KRW)
-
Includes:
50 cm µPAC Neo Plus column, ACC-VSP1006, ACC-VSP2050, ACC-GRNDC, ES993
Product Line:
µPAC Neo Plus

Learn more about your proteomics samples with Thermo Scientific™ μPAC™ Neo Plus HPLC columns. Taking your performance even further, our μPAC Neo Plus column line offers higher resolution than the previous generation of μPAC™ Neo columns, ensuring that you can extract the maximum amount of information from highly complex samples using low-flow chromatography. Column installation is simple and reliable with Thermo Scientific™ nanoViper™ Fingertight Fittings.

The μPAC Neo Plus column range is the ideal choice for a variety of proteomics experiments, including fast label-free quantitation, comprehensive identification, and single cell analysis.

Performance and Robustness

- μPAC Neo Plus columns operate at moderate pressures, allowing for longer flow paths that increase separation performance and sensitivity. Operating at lower back pressures also extends column lifetime.

Column-to-Column Reproducibility

- The unique micromachining manufacturing of our μPAC Neo Plus column line produces columns that are virtually identical. This results in increased injection-to-injection and column-to-column reproducibility, providing you with greater confidence in your results throughout the duration of a research study.

Perfect Flow Path Order

- μPAC Neo Plus columns are designed with a perfect flow path order, eliminating heterogeneous flow paths. The ordered flow path of the columns minimizes dispersion to the overall separation, resulting in sharper and more intense chromatographic peaks.

Simple column setup with starter kits

- μPAC Neo Plus Column Kits include all accessories required to run µPAC Neo Plus columns with a Thermo Scientific™ EASY-Spray™ source.

Specifications
Bed Length50 cm
Bed Width180 µm
Column FormatDouble nanoViper
Column TypeCylindrical Micro Pillar Array
EndcappedYes
Flow Rate100 to 750 nL/min
For Use WithEASY-Spray ion source
For Use With (Application)Bottom-up Proteomics
Includes50 cm µPAC Neo Plus column, ACC-VSP1006, ACC-VSP2050, ACC-GRNDC, ES993
Interpillar Distance1.25 µm
Max. Pressure6526 psi (450 bar)
Packing MaterialSilicon Chip
Pillar Diameter2.5 µm
Pillar Length16 µm
Stationary PhaseC18
Temperature60°CC
Volume (Metric)1.5 µL
Pore Size100 to 300 Å
Product LineµPAC Neo Plus
TypeµPAC Trapping Columns kit - 50 cm µPAC Neo Plus column, ACC-VSP1006, ACC-VSP2050, ACC-GRNDC, ES993
pH1.5 to 7
Unit SizeEach