Arcturus™ Paradise™ PLUS 1.5 Round Kit, no solvents - FAQs
Arcturus™ Paradise™ PLUS 1.5 Round Kit, no solvents - FAQs
View additional product information for Arcturus™ Paradise™ PLUS 1.5 Round Kit, no solvents - FAQs (KIT0311NS)
3 product FAQs found
How do I locate the overview image on the Arcturus XT LCM Instrument?
First, optimize the visualization of the image in the live video at 2X by increasing or decreasing the brightness setting, then right-click on the overview image and select "remember settings". This resets your optimized image settings, and when you select "re-acquire overview image" you will now generate a perfectly exposed overview image.
With the ArcturusXT LCM Instrument, which laser should I use to isolate my region of interest? UV cutting laser or IR capture laser?
For a few cells: The gentle IR-only approach is the best recommendation for preserving nucleic acids and verifying that the desired material is collected on the cap. For groups of cells or large tumor regions: Mount your tissue on a membrane slide and take advantage of the power of the IR, or IR + UV cutting laser to cut around your region of interest quickly and cleanly.
What is Arcturus Laser Capture Microdissection (LCM)?
The ArcturusXT LCM Instrument has a proprietary combination of a gentle IR laser and a powerful UV laser that work in conjunction to efficiently isolate cells from frozen sections or paraffin embedded sections without changing morphology or integrity of the biological content. The IR laser helps to capture the cells of interest, and the UV laser microdissects the captured cells. This is done without affecting the morphology of the cells, and allows for visual inspection of the remaining specimen to help ensure the quality of the sample collected. In addition to the flexibility that the dual laser system provides, there is the flexibility of multiple stage inserts for various sample types. A wide-slide stage format is available for neurobiology researchers working with very large sections of brain tissue. The petri dish stage insert enables live-cell microdissection applications such as stem cell research and other rare-cell isolations.