Applied Biosystems™

Turbo Labeling™ Kit, Cy3 dye

Catalog number:  KIT0609

 Related applications:

Laser Capture Microdissection

| Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Arcturus® Turbo Labeling™ Kits offer a proprietary, non-enzymatic technology optimized for the labeling of unmodified, amplified RNA (aRNA) for gene expression profiling. The unmodified aRNA is labeled post-amplification, thereby avoiding the need to incorporate modified nucleotides during RNA amplification. The use of unmodified nucleotides in the amplification process results in unmodified aRNA with higher yields and longer aRNA fragments, thus providing better representation of the mRNA transcript for downstream analysis (Figures 1 and 2).

Key product features:
• Simple – platform-independent aRNA labeling protocol typically takes less than 30 minutes
• Efficient – unlabeled aRNA can be preserved for downstream validation
• Effective – use of unmodified nucleotides permits exceptional representation of mRNA transcripts
Arcturus® Turbo Labeling™ Kits include reagents to support labeling of 12 samples with either Cy3*⁄Cy5 dyes (Figure 3) or with biotin for hybridization to cDNA or oligonucleotide arrays (Figure 4).

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Downstream Application: Microarray Analysis
Final Product: RNA (Amplified, Labeled)
Includes Label or Dye: Yes
Label or Dye: Cy™3
Labeling Method: Direct Labeling
Labeling Target: RNA (General)
Product Line: Turbo Labeling™
Product Size: 12 samples

Contents & storage

Cy3 Reagent (store at 4 to 8°C), 10X Labeling Buffer (store at 4 to 8°C), Blocking Reagent (store at -15 to -30°C), Purification Columns and Collection Tubes (store at 4 to 8°C).

Documents

Manuals & protocols