InSight Kits
El producto real puede variar
InSight Kits
InSight Kits
InSight Kits
InSight Kits
InSight Kits
InSight Kits
Thermo Scientific™
InSight Kits
Proactively monitor your cold storage equipment’s temperature, energy consumption, and other parameters using Wi-Fi or proprietary wireless protocols.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoPara utilizar con (equipo)
KITT3C1D2WIF1002Refrigeradores
KITT3C1D2WIF1001Ultracongeladores
Número de catálogo KITT3C1D2WIF1002
Precio (MXN)
-
Para utilizar con (equipo):
Refrigeradores
Thermo Scientific™ InSight is a wireless monitoring solution that has been designed to proactively monitor your cold storage equipment. InSight consists of wireless modules (motes), sensors and software tools that allow you to monitor your cold storage equipment’s temperature, energy consumption, door access, reliability, predictive failure analytics, as well as other parameters using Wi-Fi or proprietary wireless protocols. Alarms are issued automatically to users’ email and mobile devices when setpoint breaches are detected in the system.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
  • Cabinet temperature
  • Electrical use and energy consumption
  • Door openings
  • Ambient temperature
InSight Analytics monitors your cold storage equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
  • Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight gateway server. There, the server collects and transmits the data to Cloud storage.
  • As an alternative to Wi-Fi, wireless 915MHZ systems (North America only) are also available for smaller installations (25 motes or less).
  • Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the cold storage equipment's health to industry averages.
  • This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
Not available outside North America
Especificaciones
CalibraciónSin calibrar
Descripción802.11(n) for refrigerators. T3C1D2 Includes PT100, current transducer and door switch. Also includes 1 asset license; InSight refigerator license
Para utilizar con (equipo)Refrigeradores
Tipos de sensoresRTD Sensor, Thermocouple
Tipos de alarmaE-mail
Tipo de pantallaSin pantalla
Software TypeInsightlink Gateway
TipoInsight Kit
Unit SizeEach