You need to enable JavaScript to run this app.
Popular
Applications & Techniques
Shop All Products
Services
Support
Order Status
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
Error:
Error:
This product (Catalog number KNZ1161) has been discontinued. Please contact
Customer Service
for possible alternatives.
Sign in
Don't have an account ?
Create Account
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
Account
Check Order Status
Popular
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Applications & Techniques
See Links For Applications & Techniques
Brands
See Links For Brands
Thermo Scientific
Applied Biosystems
Invitrogen
Fisher Scientific
Unity Lab Services
Patheon
PPD
Life Sciences
See Links For Life Sciences
Antibodies
Cell Analysis
Cell Culture & Transfection
Cloning
DNA & RNA Extraction & Analysis
Epigenetics & ncRNA Research
Flow Cytometry
Gene Expression Analysis & Genotyping
Genome Editing
Microarray Analysis
Oligos, Primers, Probes & Genes
PCR
Protein Biology
Real-Time PCR
RNAi
Sequencing
Stem Cell Research
Synthetic Biology
Industrial & Applied Sciences
See Links For Industrial & Applied Sciences
Agrigenomics
Animal Health
Bioprocessing
Biotech Company Solutions
Cement, Coal & Minerals
Chromatography
Drug Discovery & Development
Electron Microscopy
Environmental
Food & Beverage
Forensics
Geosciences
Manufacturing & Processing
Mass Spectrometry
Materials Science
Microbiology
Pharma & Biopharma
Radiation Detection & Measurement
Safety & Security Threat Detection
Semiconductor Analysis
Spectroscopy, Elemental & Isotope Analysis
Clinical & Diagnostics
See Links For Clinical & Diagnostics
Allergy Diagnostics
Anatomical Pathology
Autoimmune Diagnostics
Biobanking
Cancer Research
Cell & Gene Therapy
Clinical Genomics
Clinical Mass Spectrometry
Clinical Microbiology
Clinical & Translational Research
Diagnostic Development
Diagnostic Testing
Precision Medicine
Preclinical to Companion Diagnostic Development
Public Health
Lab Solutions
See Links For Lab Solutions
Chemicals
Lab Equipment
Lab Plasticware & Supplies
Sample Storage & Management
Digital Solutions
See Links For Digital Solutions
Lab Informatics
Connect Platform
Instrument Connectivity
Apps & Analysis Software
Lab Automation
Digital Services
Digital Commerce
Shop All Products
See Links For Shop All Products
Most Popular Products
Pipettes & Pipette Tips
Beakers
Nalgene Labware
Lab Centrifuges
Ultra Low–Temperature Freezers
Lipofectamine Reagents
Oligos, Primers, Probes & Genes
SuperScript Reverse Transcriptase
TaqMan Real-Time PCR Assays
TRIzol Reagents
Antibodies
ELISA Kits
Most Popular Categories
Microplates
PCR Equipment and Supplies
Molecular Biology Reagents and Kits
Chromatography Columns and Cartridges
Chromatography Supplies
Cell Culture Media
Microbiological Media and Media Additives
Antibiotics and Supplements
Cell Analysis Products
Gel Electrophoresis Equipment and Supplies
Western Blot Products
Microplate Readers and Accessories
Spectroscopy
Order Tools
How to Order
Quick Order
Track Your Order
Order History
Order Support
Contact Us
Shop All Products
See Links For Shop All Products
Product Selection Guides
New Products
Promotions
Online Offers
Services
See Links For Services
Custom Services
Enterprise-Level Lab Informatics
Enterprise Services
Financial & Leasing Solutions
Instrument Services
OEM & Commercial Supply
Training Services
CDMO & Clinical Trial Services
Support
See Links For Support
Product Documentation
Manuals
Safety Data Sheets (SDS)
Certificates of Analysis & Conformance
CE Declarations of Conformity
RoHS Certificates
Manufacturing Site ISO Certifications
Other Product Information
Citations & References
Product Literature
Vector Maps
Spectral Data
Media Formulations
Product Support
Knowledge Base & FAQs
Instrument Support
Support Centers
Contact Us
Educational Resources
Learning Centers
Connect to Science
Digital Transformation
Gibco Cell Culture Basics
Protein Methods Library
Supplemental Protocols
Newsletters & Journals
Training Services
Events
Popular Tools & Calculators
Lab Apps for Mobile Devices
Oligos Tools & Utilities Hub
Tm Calculator for PCR
Molecular Biology Tools
Gibco Media Formulation Tool
Cell Culture Useful Numbers
Fluorescence SpectraViewer
Flow Cytometry Panel Builder
Cell Staining Tool
Protein Gel Conversion Tool
Switch to Nunc Plastics Tool
Transfection Protocol Calculator